Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576048_m1
          See other HTR7 GE Assays ›
          Gene Symbol:
          Htr7 LOC103694905
          Gene Name:
          5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled
          5-hydroxytryptamine receptor 7
          Gene Aliases:
          5Ht7
          -
          Chromosome Location:
          Chr.1: 254547964 - 254671811 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906944: 278684 - 399514 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00576048_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htr7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022938.2 NP_075227.1 1 - 2 655 85
          XM_006231307.2 XP_006231369.1 1 - 2 870 85
          GenBank mRNA L22558.1 - 1 - 2 655 85
          U68489.1 - 1 - 2 655 85
          U68490.1 - 2 - 3 655 85
          Gene Symbol: LOC103694905
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773900.1 XP_008772122.1 870 85
          XM_008773901.1 XP_008772123.1 870 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htr7
          Click here to find siRNAs for Gene LOC103694905

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          smooth muscle contraction
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-activating serotonin receptor signaling pathway
          serotonin receptor signaling pathway
          chemical synaptic transmission
          memory
          circadian rhythm
          positive regulation of neuron projection development
          negative regulation of serotonin secretion
          urinary bladder smooth muscle contraction
          vasoconstriction
          negative regulation of circadian sleep/wake cycle, REM sleep
          behavioral response to pain
          detection of mechanical stimulus involved in sensory perception of pain
          response to corticosterone
          response to electrical stimulus
          micturition
          long-term synaptic potentiation
          dopaminergic neuron differentiation
          G-protein coupled serotonin receptor activity
          neurotransmitter receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P128139 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P371317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31451 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128341 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73327 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159367 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P371317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P371317 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P128139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P371317 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P371317 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P412178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90805 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67420 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47162 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61707 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10942 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9700 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56993 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729830 - -
          Affymetrix Rat Gene 1.0 ST Array 10729831 - -
          Affymetrix Rat Gene 1.0 ST Array 10729832 - -
          Affymetrix Rat Gene 1.0 ST Array 10729826 - -
          Affymetrix Rat Gene 1.0 ST Array 10729827 - -
          Affymetrix Rat Gene 1.0 ST Array 10729833 - -
          Affymetrix Rat Gene 1.1 ST Array 10729825 - -
          Rat Genome 230 2.0 Array 1387466_a_at - -
          Rat Genome 230 2.0 Array 1387466_PM_a_at - -
          Rat Genome 230 2.0 Array 1369119_PM_a_at - -
          Rat Genome 230 2.0 Array 1369119_a_at - -
          Rat Genome 230 2.0 Array 1371290_PM_a_at - -
          Rat Genome 230 2.0 Array 1371290_a_at - -
          Affymetrix HT Rat Focus Array 1387466_a_at - -
          Affymetrix HT Rat Focus Array 1371290_a_at - -
          Affymetrix HT Rat Focus Array 1369119_a_at - -
          Affymetrix HT Rat Focus Array 1369119_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387466_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371290_PM_a_at - -
          Rat Expression Set 230 1387466_a_at - -
          Rat Expression Set 230 1371290_PM_a_at - -
          Rat Expression Set 230 1387466_PM_a_at - -
          Rat Expression Set 230 1369119_PM_a_at - -
          Rat Expression Set 230 1369119_a_at - -
          Rat Expression Set 230 1371290_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729825 - -
          Rat Genome U34 Set L22558_at - -
          Rat Genome U34 Set U68491_at - -
          Rat Genome U34 Set U68491_at - -
          Rat Genome U34 Set L22558_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20858674 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367458 - -
          RatRef-12 Expression BeadChip ILMN_64336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline