Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576617_m1
          See other MUG1 GE Assays ›
          Gene Symbol:
          Mug1 A1i3 LOC100911784
          Gene Name:
          murinoglobulin 1
          alpha-1-inhibitor III
          murinoglobulin-1-like
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.4: 154676387 - 154729288 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 154505735 - 154565099 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 154599966 - 154637598 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn00576617_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mug1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023103.1 NP_075591.1 4 - 5 532 106
          XM_006237282.2 XP_006237344.1 4 - 5 572 106
          GenBank mRNA M22360.1 - 3 - 4 246 106
          X52984.1 - 4 - 5 532 106
          Gene Symbol: A1i3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037975.2 NP_001033064.2 4 - 5 560 106
          XM_006237281.2 XP_006237343.1 4 - 5 572 106
          GenBank mRNA J03552.1 - 4 - 5 560 106
          Gene Symbol: LOC100911784
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763278.1 XP_008761500.1 2 - 3 244 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mug1
          Click here to find siRNAs for Gene A1i3
          Click here to find siRNAs for Gene LOC100911784

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          acute-phase response
          inflammatory response
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity
          endopeptidase inhibitor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0400012458.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012466.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012456.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012461.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029572.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029574.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029578.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029580.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012463.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029577.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012465.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029571.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012462.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029581.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029582.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012457.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029570.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029573.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029579.rn.1 TC0400004147.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012459.rn.1 TC0400001716.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012464.rn.1 TC0400001716.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline