Search
Search

Gucy1a2| Gene Symbol: Gucy1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023956.1 | NP_076446.1 | 1 - 2 | 720 | 76 | |
| GenBank mRNA | AB096080.1 | - | 1 - 2 | 720 | 76 | |
| AB097860.1 | - | 1 - 2 | 300 | 76 | ||
| AF109963.2 | - | 1 - 2 | 720 | 76 | ||
Set Membership: |
|