Search
Search
| Gene Symbol: Grm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031040.1 | NP_112302.1 | 5 - 6 | 1567 | 96 | |
| GenBank mRNA | D16817.1 | - | 5 - 6 | 1567 | 96 | |
| U06832.1 | - | 5 - 6 | 1302 | 96 | ||
Set Membership: |
|