Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579856_m1
          See other SHARPIN GE Assays ›
          Gene Symbol:
          Sharpin
          Gene Name:
          SHANK-associated RH domain interactor
          Gene Aliases:
          Conneck1
          Chromosome Location:
          Chr.7: 117413151 - 117417363 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579856_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sharpin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031153.2 NP_112415.1 2 - 3 719 95
          GenBank mRNA AF203906.1 - 2 - 3 393 95
          BC083553.1 - 2 - 3 719 7189111 95
          FQ213170.1 - 2 - 3 337 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003391.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sharpin

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrion organization
          brain development
          epidermis development
          regulation of tumor necrosis factor-mediated signaling pathway
          apoptotic nuclear changes
          keratinization
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of inflammatory response
          protein homooligomerization
          protein linear polyubiquitination
          regulation of CD40 signaling pathway
          protein binding
          zinc ion binding
          polyubiquitin binding
          protein complex binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1011726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42354 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011726 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1011726 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1011726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88815 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10904913 - -
          Affymetrix Rat Gene 1.0 ST Array 10904912 - -
          Affymetrix Rat Gene 1.0 ST Array 10904911 - -
          Affymetrix Rat Gene 1.0 ST Array 10904917 - -
          Affymetrix Rat Gene 1.0 ST Array 10904914 - -
          Affymetrix Rat Gene 1.0 ST Array 10904919 - -
          Affymetrix Rat Gene 1.0 ST Array 10904920 - -
          Affymetrix Rat Gene 1.0 ST Array 10904915 - -
          Affymetrix Rat Gene 1.0 ST Array 10904916 - -
          Affymetrix Rat Gene 1.0 ST Array 10904918 - -
          Affymetrix Rat Gene 1.1 ST Array 10904910 - -
          Rat Genome 230 2.0 Array 1368069_PM_at - -
          Rat Genome 230 2.0 Array 1368069_at - -
          Affymetrix HT Rat Focus Array 1368069_PM_at - -
          Affymetrix HT Rat Focus Array 1368069_at - -
          Affymetrix Rat Clariom S Assay PSR0700026330.rn.1 TC0700003391.rn.1 RTA TC
          Rat Expression Set 230 1368069_at - -
          Rat Expression Set 230 1368069_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10904910 - -
          Rat Genome U34 Set rc_AA858478_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21957524 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373042 - -
          RatRef-12 Expression BeadChip ILMN_58452 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline