Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580926_m1
          See other SLC37A4 GE Assays ›
          Gene Symbol:
          Slc37a4
          Gene Name:
          solute carrier family 37 (glucose-6-phosphate transporter), member 4
          Gene Aliases:
          G6pt1
          Chromosome Location:
          Chr.8: 48716939 - 48723024 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580926_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc37a4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031589.2 NP_113777.2 2 - 3 427 87
          XM_008766152.1 XP_008764374.1 1 - 2 253 87
          GenBank mRNA AF080468.1 - 1 - 2 257 87
          BC070889.1 - 2 - 3 427 7098680 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000567.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc37a4

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neutrophil homeostasis
          cytokine production
          myeloid progenitor cell differentiation
          glycogen metabolic process
          lactate metabolic process
          triglyceride metabolic process
          cellular calcium ion homeostasis
          response to nutrient
          steroid metabolic process
          response to glucose
          response to organic cyclic compound
          glucose-6-phosphate transport
          neutrophil chemotaxis
          negative regulation of chemokine production
          post-embryonic hemopoiesis
          glucose homeostasis
          cholesterol homeostasis
          positive regulation of catalytic activity
          respiratory burst
          transmembrane transport
          glucose-6-phosphate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1023264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55354 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023264 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023264 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1023264 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12602 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1023264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102078 - -
          Agilent Rat cDNA Microarray (G4105A) 700606687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909520 - -
          Affymetrix Rat Gene 1.0 ST Array 10909510 - -
          Affymetrix Rat Gene 1.0 ST Array 10909514 - -
          Affymetrix Rat Gene 1.0 ST Array 10909519 - -
          Affymetrix Rat Gene 1.0 ST Array 10909511 - -
          Affymetrix Rat Gene 1.0 ST Array 10909516 - -
          Affymetrix Rat Gene 1.0 ST Array 10909513 - -
          Affymetrix Rat Gene 1.0 ST Array 10909512 - -
          Affymetrix Rat Gene 1.0 ST Array 10909515 - -
          Affymetrix Rat Gene 1.0 ST Array 10909517 - -
          Affymetrix Rat Gene 1.1 ST Array 10909509 - -
          Rat Genome 230 2.0 Array 1386960_PM_at - -
          Rat Genome 230 2.0 Array 1386960_at - -
          Affymetrix HT Rat Focus Array 1386960_at - -
          Affymetrix HT Rat Focus Array 1386960_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800003575.rn.1 TC0800000567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003566.rn.1 TC0800000567.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003577.rn.1 TC0800000567.rn.1 RTA TC
          Rat Expression Set 230 1386960_at - -
          Rat Expression Set 230 1386960_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909509 - -
          Rat Genome U34 Set AF080468_g_at - -
          Rat Genome U34 Set AF080468_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22146943 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359078 - -
          RatRef-12 Expression BeadChip ILMN_51888 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline