Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582748_m1
          See other CTCF GE Assays ›
          Gene Symbol:
          Ctcf
          Gene Name:
          CCCTC-binding factor (zinc finger protein)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 37600377 - 37649674 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582748_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctcf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031824.1 NP_114012.1 10 - 11 2007 105
          XM_008772537.1 XP_008770759.1 8 - 9 1974 105
          XM_008772539.1 XP_008770761.1 8 - 9 1974 105
          GenBank mRNA AF133731.2 - 10 - 11 2043 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000485.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctcf

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          chromosome segregation
          maintenance of DNA methylation
          positive regulation of gene expression
          negative regulation of gene expression
          chromatin modification
          nucleosome positioning
          regulation of histone methylation
          regulation of histone acetylation
          regulation of gene expression, epigenetic
          regulation of molecular function, epigenetic
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          chromatin insulator sequence binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P630249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828801 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180150 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P630249 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P630249 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P630249 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P630249 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701337620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1303 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55388 - -
          Agilent Rat cDNA Microarray (G4105A) 701337620 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807305 - -
          Affymetrix Rat Gene 1.0 ST Array 10807307 - -
          Affymetrix Rat Gene 1.0 ST Array 10807308 - -
          Affymetrix Rat Gene 1.0 ST Array 10807301 - -
          Affymetrix Rat Gene 1.0 ST Array 10807302 - -
          Affymetrix Rat Gene 1.0 ST Array 10807303 - -
          Affymetrix Rat Gene 1.0 ST Array 10807309 - -
          Affymetrix Rat Gene 1.0 ST Array 10807310 - -
          Affymetrix Rat Gene 1.0 ST Array 10807304 - -
          Affymetrix Rat Gene 1.0 ST Array 10807306 - -
          Affymetrix Rat Gene 1.1 ST Array 10807300 - -
          Rat Genome 230 2.0 Array 1370337_PM_at - -
          Rat Genome 230 2.0 Array 1370337_at - -
          Affymetrix HT Rat Focus Array 1370337_at - -
          Affymetrix HT Rat Focus Array 1370337_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900004055.rn.1 TC1900000485.rn.1 RTA TC
          Rat Expression Set 230 1370337_PM_at - -
          Rat Expression Set 230 1370337_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807300 - -
          Rat Genome U34 Set rc_AI030850_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21920679 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370286 - -
          RatRef-12 Expression BeadChip ILMN_49797 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline