Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583768_m1
          See other ZFP354A GE Assays ›
          Gene Symbol:
          Zfp354a
          Gene Name:
          zinc finger protein 354A
          Gene Aliases:
          Kid1, Tcf17, Znf354a
          Chromosome Location:
          Chr.10: 36587094 - 36600172 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583768_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp354a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_052798.2 NP_434685.1 2 - 3 470 77
          XM_006246227.2 XP_006246289.1 3 - 4 1617 77
          GenBank mRNA M96548.1 - 2 - 3 472 77

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp354a

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to hypoxia
          kidney development
          transcription, DNA-templated
          nucleolar fragmentation
          response to folic acid
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P101608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117104 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167237 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101608 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101608 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P101608 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12741 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700327476 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1998 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51969 - -
          Agilent Rat cDNA Microarray (G4105A) 700327476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733313 - -
          Affymetrix Rat Gene 1.0 ST Array 10733311 - -
          Affymetrix Rat Gene 1.0 ST Array 10733316 - -
          Affymetrix Rat Gene 1.0 ST Array 10733317 - -
          Affymetrix Rat Gene 1.0 ST Array 10733315 - -
          Affymetrix Rat Gene 1.0 ST Array 10733312 - -
          Affymetrix Rat Gene 1.0 ST Array 10733314 - -
          Affymetrix Rat Gene 1.1 ST Array 10733310 - -
          Rat Genome 230 2.0 Array 1368877_at - -
          Rat Genome 230 2.0 Array 1368877_PM_at - -
          Affymetrix HT Rat Focus Array 1368877_PM_at - -
          Affymetrix HT Rat Focus Array 1368877_at - -
          Rat Expression Set 230 1368877_PM_at - -
          Rat Expression Set 230 1368877_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733310 - -
          Rat Genome U34 Set M96548_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21158632 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371674 - -
          RatRef-12 Expression BeadChip ILMN_56707 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline