Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583895_m1
          See other CCNH GE Assays ›
          Gene Symbol:
          Ccnh
          Gene Name:
          cyclin H
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 13593100 - 13613910 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583895_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccnh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_052981.2 NP_443213.2 9 - 10 1244 96
          GenBank mRNA AF154914.1 - 8 - 9 941 96
          AJ276495.1 - 9 - 10 1244 96
          BC059109.1 - 8 - 9 1047 6886953 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000103.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccnh

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          transcription from RNA polymerase II promoter
          protein phosphorylation
          biological_process
          phosphorylation
          positive regulation of transcription from RNA polymerase II promoter
          molecular_function
          DNA-dependent ATPase activity
          RNA polymerase II carboxy-terminal domain kinase activity
          kinase activity
          cyclin-dependent protein serine/threonine kinase regulator activity
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P400355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109866 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P400355 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P400355 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P400355 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P400355 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700526935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42715 - -
          Agilent Rat cDNA Microarray (G4105A) 700526935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812365 - -
          Affymetrix Rat Gene 1.0 ST Array 10812369 - -
          Affymetrix Rat Gene 1.0 ST Array 10812368 - -
          Affymetrix Rat Gene 1.0 ST Array 10812373 - -
          Affymetrix Rat Gene 1.0 ST Array 10812370 - -
          Affymetrix Rat Gene 1.0 ST Array 10812374 - -
          Affymetrix Rat Gene 1.0 ST Array 10812366 - -
          Affymetrix Rat Gene 1.0 ST Array 10812371 - -
          Affymetrix Rat Gene 1.0 ST Array 10812364 - -
          Affymetrix Rat Gene 1.0 ST Array 10812372 - -
          Affymetrix Rat Gene 1.0 ST Array 10812367 - -
          Affymetrix Rat Gene 1.1 ST Array 10812363 - -
          Rat Genome 230 2.0 Array 1368083_at - -
          Rat Genome 230 2.0 Array 1368083_PM_at - -
          Affymetrix HT Rat Focus Array 1368083_at - -
          Affymetrix HT Rat Focus Array 1368083_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200000525.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000531.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000529.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000524.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000534.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000528.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000527.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000532.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000526.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000522.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000533.rn.1 TC0200000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000523.rn.1 TC0200000103.rn.1 RTA TC
          Rat Expression Set 230 1368083_at - -
          Rat Expression Set 230 1368083_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812363 - -
          Rat Genome U34 Set rc_AI177687_at - -
          Rat Genome U34 Set rc_AI111344_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22195724 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373618 - -
          RatRef-12 Expression BeadChip ILMN_58817 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline