Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583935_m1
          See other ADORA2A GE Assays ›
          Gene Symbol:
          Adora2a
          Gene Name:
          adenosine A2a receptor
          Gene Aliases:
          A2ar, ADENO, Adora2l1
          Chromosome Location:
          Chr.20: 14265252 - 14282873 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583935_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adora2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053294.4 NP_445746.3 2 - 3 752 58
          GenBank mRNA AF115508.1 - 1 - 2 258 58
          AF228684.1 - 1 - 2 358 58
          BC081727.1 - 2 - 3 752 7121208 58
          L08102.1 - 1 - 2 324 58
          M91214.2 - 1 - 2 609 58
          S47609.1 - 1 - 2 398 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001364.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adora2a

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          synaptic transmission, dopaminergic
          adenosine receptor signaling pathway
          response to amphetamine
          regulation of transcription, DNA-templated
          negative regulation of protein kinase activity
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          protein kinase C-activating G-protein coupled receptor signaling pathway
          synaptic transmission, cholinergic
          locomotory behavior
          negative regulation of cell proliferation
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          positive regulation of glutamate secretion
          positive regulation of acetylcholine secretion, neurotransmission
          regulation of norepinephrine secretion
          positive regulation of cAMP biosynthetic process
          response to caffeine
          positive regulation of synaptic transmission, GABAergic
          synaptic transmission, glutamatergic
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          negative regulation of locomotion
          vasodilation
          response to drug
          eating behavior
          negative regulation of vascular permeability
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to alkaloid
          negative regulation of neuron apoptotic process
          positive regulation of circadian sleep/wake cycle, sleep
          negative regulation of alpha-beta T cell activation
          astrocyte activation
          regulation of synaptic plasticity
          neuron projection morphogenesis
          positive regulation of protein secretion
          negative regulation of inflammatory response
          regulation of mitochondrial membrane potential
          membrane depolarization
          regulation of calcium ion transport
          positive regulation of synaptic transmission, glutamatergic
          excitatory postsynaptic potential
          inhibitory postsynaptic potential
          prepulse inhibition
          positive regulation of apoptotic signaling pathway
          G-protein coupled adenosine receptor activity
          protein binding
          enzyme binding
          type 5 metabotropic glutamate receptor binding
          identical protein binding
          protein heterodimerization activity
          alpha-actinin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1034950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034950 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034950 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1034950 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P664121 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19648 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58435 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10832578 - -
          Affymetrix Rat Gene 1.0 ST Array 10832582 - -
          Affymetrix Rat Gene 1.0 ST Array 10832579 - -
          Affymetrix Rat Gene 1.0 ST Array 10832581 - -
          Affymetrix Rat Gene 1.0 ST Array 10832583 - -
          Affymetrix Rat Gene 1.0 ST Array 10832580 - -
          Affymetrix Rat Gene 1.1 ST Array 10832577 - -
          Rat Genome 230 2.0 Array 1368301_at - -
          Rat Genome 230 2.0 Array 1368300_PM_at - -
          Rat Genome 230 2.0 Array 1368300_at - -
          Rat Genome 230 2.0 Array 1368301_PM_at - -
          Affymetrix HT Rat Focus Array 1368300_at - -
          Affymetrix HT Rat Focus Array 1368300_PM_at - -
          Affymetrix HT Rat Focus Array 1368301_at - -
          Affymetrix HT Rat Focus Array 1368301_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000011266.rn.1 TC2000001364.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000011267.rn.1 TC2000001364.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000011265.rn.1 TC2000001364.rn.1 RTA TC
          Rat Expression Set 230 1368300_PM_at - -
          Rat Expression Set 230 1368301_PM_at - -
          Rat Expression Set 230 1368300_at - -
          Rat Expression Set 230 1368301_at - -
          Affymetrix Rat Gene 1.0 ST Array 10832577 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22387539 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364931 - -
          RatRef-12 Expression BeadChip ILMN_58181 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline