Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584313_m1
          See other FETUB GE Assays ›
          Gene Symbol:
          Fetub
          Gene Name:
          fetuin B
          Gene Aliases:
          Fet, Pp63
          Chromosome Location:
          Chr.11: 81648890 - 81661915 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584313_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fetub
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053348.1 NP_445800.1 3 - 4 541 94
          XM_006248549.1 XP_006248611.1 4 - 5 495 94
          XM_006248550.1 XP_006248612.1 4 - 5 858 94
          GenBank mRNA AJ242926.1 - 3 - 4 541 94
          BC070508.1 - 4 - 5 480 7099154 94
          FQ209398.1 - 3 - 4 552 94
          FQ209449.1 - 4 - 5 476 94
          FQ209518.1 - 3 - 4 492 94
          FQ209610.1 - 3 - 4 527 94
          FQ209832.1 - 3 - 4 492 94
          FQ210294.1 - 3 - 4 492 94
          FQ210335.1 - 3 - 4 493 94
          FQ211050.1 - 3 - 4 492 94
          FQ211248.1 - 3 - 4 527 94
          FQ218476.1 - 3 - 4 523 94
          FQ218557.1 - 4 - 5 471 94
          FQ218709.1 - 3 - 4 492 94
          FQ218884.1 - 4 - 5 463 94
          FQ218905.1 - 3 - 4 492 94
          FQ218930.1 - 3 - 4 525 94
          FQ218949.1 - 3 - 4 342 94
          FQ219048.1 - 3 - 4 523 94
          FQ219152.1 - 3 - 4 529 94
          FQ219190.1 - 3 - 4 527 94
          FQ219266.1 - 3 - 4 492 94
          FQ219352.1 - 3 - 4 492 94
          FQ219426.1 - 1 - 2 94 94
          FQ219454.1 - 3 - 4 492 94
          FQ219459.1 - 3 - 4 490 94
          FQ219500.1 - 3 - 4 495 94
          FQ219513.1 - 3 - 4 564 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001889.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fetub

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          single fertilization
          binding of sperm to zona pellucida
          negative regulation of endopeptidase activity
          cysteine-type endopeptidase inhibitor activity
          metalloendopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P379377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379377 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379377 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P379377 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P379377 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700305867 - -
          Agilent Rat cDNA Microarray (G4105A) 700305867 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755149 - -
          Affymetrix Rat Gene 1.0 ST Array 10755153 - -
          Affymetrix Rat Gene 1.0 ST Array 10755150 - -
          Affymetrix Rat Gene 1.0 ST Array 10755152 - -
          Affymetrix Rat Gene 1.0 ST Array 10755155 - -
          Affymetrix Rat Gene 1.0 ST Array 10755151 - -
          Affymetrix Rat Gene 1.0 ST Array 10755154 - -
          Affymetrix Rat Gene 1.0 ST Array 10755156 - -
          Affymetrix Rat Gene 1.1 ST Array 10755148 - -
          Rat Genome 230 2.0 Array 1387082_at - -
          Rat Genome 230 2.0 Array 1387082_PM_at - -
          Affymetrix HT Rat Focus Array 1387082_at - -
          Affymetrix HT Rat Focus Array 1387082_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100012060.rn.1 TC1100001889.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100012059.rn.1 TC1100001889.rn.1 RTA TC
          Rat Expression Set 230 1387082_PM_at - -
          Rat Expression Set 230 1387082_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755148 - -
          Rat Genome U34 Set rc_AI169740_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21609559 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372913 - -
          RatRef-12 Expression BeadChip ILMN_50667 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline