Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584780_m1
          See other SLC17A6 GE Assays ›
          Gene Symbol:
          Slc17a6
          Gene Name:
          solute carrier family 17 (vesicular glutamate transporter), member 6
          Gene Aliases:
          Dnpi, Vglut2
          Chromosome Location:
          Chr.1: 106998704 - 107038717 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584780_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc17a6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053427.1 NP_445879.1 1 - 2 547 141
          GenBank mRNA AF271235.1 - 1 - 2 547 141

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF271235.1, NM_053427.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc17a6

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          sodium ion transport
          L-glutamate transmembrane transport
          L-glutamate transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P511118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P511118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P511118 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P511118 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P511118 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700370181 - -
          Agilent Rat cDNA Microarray (G4105A) 700370181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10707329 - -
          Affymetrix Rat Gene 1.0 ST Array 10707327 - -
          Affymetrix Rat Gene 1.0 ST Array 10707326 - -
          Affymetrix Rat Gene 1.0 ST Array 10707331 - -
          Affymetrix Rat Gene 1.0 ST Array 10707330 - -
          Affymetrix Rat Gene 1.0 ST Array 10707328 - -
          Affymetrix Rat Gene 1.0 ST Array 10707333 - -
          Affymetrix Rat Gene 1.0 ST Array 10707336 - -
          Affymetrix Rat Gene 1.0 ST Array 10707335 - -
          Affymetrix Rat Gene 1.0 ST Array 10707334 - -
          Affymetrix Rat Gene 1.0 ST Array 10707337 - -
          Affymetrix Rat Gene 1.0 ST Array 10707332 - -
          Affymetrix Rat Gene 1.1 ST Array 10707325 - -
          Rat Genome 230 2.0 Array 1368564_at - -
          Rat Genome 230 2.0 Array 1368564_PM_at - -
          Affymetrix HT Rat Focus Array 1368564_PM_at - -
          Affymetrix HT Rat Focus Array 1368564_at - -
          Affymetrix Rat Clariom S Assay PSR0100012707.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012708.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012713.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012714.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012716.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012705.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012712.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012703.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012717.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012706.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012709.rn.1 TC0100001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012715.rn.1 TC0100001569.rn.1 RTA TC
          Rat Expression Set 230 1368564_PM_at - -
          Rat Expression Set 230 1368564_at - -
          Affymetrix Rat Gene 1.0 ST Array 10707325 - -
          Rat Genome U34 Set rc_AI060126_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21603659 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348843 - -
          RatRef-12 Expression BeadChip ILMN_59963 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline