Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586242_m1
          See other ST14 GE Assays ›
          Gene Symbol:
          St14
          Gene Name:
          suppression of tumorigenicity 14 (colon carcinoma)
          Gene Aliases:
          MT-SP1
          Chromosome Location:
          Chr.8: 32240113 - 32280813 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586242_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: St14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053635.2 NP_446087.1 1 - 2 204 58
          GenBank mRNA AB037898.1 - 1 - 2 78 58
          AB049189.1 - 1 - 2 204 58
          BC097271.1 - 1 - 2 109 7445508 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002176.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene St14

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          proteolysis
          keratinocyte differentiation
          epithelial cell morphogenesis involved in placental branching
          serine-type endopeptidase activity
          serine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P087801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66632 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087801 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P087801 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12878 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700311382 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700596936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90972 - -
          Agilent Rat cDNA Microarray (G4105A) 700311382 - -
          Agilent Rat cDNA Microarray (G4105A) 700596936 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915949 - -
          Affymetrix Rat Gene 1.0 ST Array 10915959 - -
          Affymetrix Rat Gene 1.0 ST Array 10915961 - -
          Affymetrix Rat Gene 1.0 ST Array 10915953 - -
          Affymetrix Rat Gene 1.0 ST Array 10915962 - -
          Affymetrix Rat Gene 1.0 ST Array 10915958 - -
          Affymetrix Rat Gene 1.0 ST Array 10915951 - -
          Affymetrix Rat Gene 1.0 ST Array 10915957 - -
          Affymetrix Rat Gene 1.0 ST Array 10915950 - -
          Affymetrix Rat Gene 1.0 ST Array 10915954 - -
          Affymetrix Rat Gene 1.0 ST Array 10915955 - -
          Affymetrix Rat Gene 1.0 ST Array 10915956 - -
          Affymetrix Rat Gene 1.0 ST Array 10915948 - -
          Affymetrix Rat Gene 1.0 ST Array 10915960 - -
          Affymetrix Rat Gene 1.0 ST Array 10915963 - -
          Affymetrix Rat Gene 1.0 ST Array 10915947 - -
          Affymetrix Rat Gene 1.0 ST Array 10915952 - -
          Affymetrix Rat Gene 1.0 ST Array 10915965 - -
          Affymetrix Rat Gene 1.0 ST Array 10915964 - -
          Affymetrix Rat Gene 1.1 ST Array 10915946 - -
          Rat Genome 230 2.0 Array 1387195_at - -
          Rat Genome 230 2.0 Array 1379968_PM_at - -
          Rat Genome 230 2.0 Array 1387195_PM_at - -
          Rat Genome 230 2.0 Array 1379968_at - -
          Affymetrix HT Rat Focus Array 1387195_PM_at - -
          Affymetrix HT Rat Focus Array 1387195_at - -
          Affymetrix Rat Clariom S Assay PSR0800017572.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017577.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017589.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017580.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017583.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017588.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017574.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017571.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017581.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017573.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017569.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017575.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017576.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017585.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017586.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017570.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017578.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017579.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017582.rn.1 TC0800002176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017584.rn.1 TC0800002176.rn.1 RTA TC
          Rat Expression Set 230 1387195_PM_at - -
          Rat Expression Set 230 1387195_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915946 - -
          Rat Genome U34 Set rc_AI030968_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22137286 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364958 - -
          RatRef-12 Expression BeadChip ILMN_55424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline