Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586650_m1
          See other YME1L1 GE Assays ›
          Gene Symbol:
          Yme1l1 LOC100910779
          Gene Name:
          YME1-like 1 ATPase
          ATP-dependent zinc metalloprotease YME1L1-like
          Gene Aliases:
          FtsH1
          -
          Chromosome Location:
          Chr.17: 89799397 - 89839452 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 89701899 - 89741953 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00586650_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Yme1l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053682.2 NP_446134.2 9 - 10 1112 63
          XM_006254339.2 XP_006254401.1 9 - 10 1133 63
          GenBank mRNA AF151784.1 - 9 - 10 976 63
          BC081751.1 - 9 - 10 1112 7108995 63
          Gene Symbol: LOC100910779
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751732.3 XP_003751780.1 1350 63
          XM_006254369.2 XP_006254431.1 1133 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Yme1l1
          Click here to find siRNAs for Gene LOC100910779

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          misfolded or incompletely synthesized protein catabolic process
          mitochondrion organization
          cell proliferation
          metalloendopeptidase activity
          ATP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P278688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173276 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P278688 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P278688 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P278688 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12896 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P278688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54971 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76382 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3268 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88852 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796815 - -
          Affymetrix Rat Gene 1.0 ST Array 10796803 - -
          Affymetrix Rat Gene 1.0 ST Array 10796805 - -
          Affymetrix Rat Gene 1.0 ST Array 10796808 - -
          Affymetrix Rat Gene 1.0 ST Array 10796814 - -
          Affymetrix Rat Gene 1.0 ST Array 10796817 - -
          Affymetrix Rat Gene 1.0 ST Array 10796816 - -
          Affymetrix Rat Gene 1.0 ST Array 10796809 - -
          Affymetrix Rat Gene 1.0 ST Array 10796806 - -
          Affymetrix Rat Gene 1.0 ST Array 10796804 - -
          Affymetrix Rat Gene 1.0 ST Array 10796807 - -
          Affymetrix Rat Gene 1.0 ST Array 10796811 - -
          Affymetrix Rat Gene 1.0 ST Array 10796818 - -
          Affymetrix Rat Gene 1.0 ST Array 10796810 - -
          Affymetrix Rat Gene 1.0 ST Array 10796813 - -
          Affymetrix Rat Gene 1.0 ST Array 10796812 - -
          Affymetrix Rat Gene 1.0 ST Array 10796801 - -
          Affymetrix Rat Gene 1.0 ST Array 10796802 - -
          Affymetrix Rat Gene 1.0 ST Array 10796819 - -
          Affymetrix Rat Gene 1.1 ST Array 10796800 - -
          Rat Genome 230 2.0 Array 1375841_PM_at - -
          Rat Genome 230 2.0 Array 1375841_at - -
          Rat Genome 230 2.0 Array 1385999_PM_at - -
          Rat Genome 230 2.0 Array 1385999_at - -
          Rat Genome 230 2.0 Array 1368843_PM_at - -
          Rat Genome 230 2.0 Array 1394059_s_at - -
          Rat Genome 230 2.0 Array 1368843_at - -
          Rat Genome 230 2.0 Array 1394059_PM_s_at - -
          Affymetrix HT Rat Focus Array 1368843_at - -
          Affymetrix HT Rat Focus Array 1394059_s_at - -
          Affymetrix HT Rat Focus Array 1394059_PM_s_at - -
          Affymetrix HT Rat Focus Array 1385999_PM_at - -
          Affymetrix HT Rat Focus Array 1368843_PM_at - -
          Affymetrix HT Rat Focus Array 1385999_at - -
          Affymetrix Rat Clariom S Assay PSR1700015770.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015773.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015788.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015767.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015768.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015772.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015777.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015778.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015803.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015760.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015763.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015776.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015765.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015775.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015779.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015792.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015793.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015795.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015797.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015761.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015764.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015766.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015769.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015791.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015794.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015796.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015798.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015799.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015802.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015762.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015789.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015801.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015771.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015780.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015781.rn.1 TC1700002356.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015800.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015804.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015805.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015806.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015787.rn.1 TC1700002357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015790.rn.1 TC1700002357.rn.1 RTA TC
          Rat Expression Set 230 1368843_at - -
          Rat Expression Set 230 1368843_PM_at - -
          Rat Expression Set 230 1394059_PM_s_at - -
          Rat Expression Set 230 1394059_s_at - -
          Rat Expression Set 230 1385999_at - -
          Rat Expression Set 230 1385999_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10796800 - -
          Rat Genome U34 Set rc_AA817882_at - -
          Rat Genome U34 Set rc_AI137819_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21709927 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349005 - -
          RatRef-12 Expression BeadChip ILMN_69823 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline