Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00586792_m1
          See other ANKH GE Assays ›
          Gene Symbol:
          Ankh
          Gene Name:
          ANKH inorganic pyrophosphate transport regulator
          Gene Aliases:
          Ank
          Chromosome Location:
          Chr.2: 80131563 - 80256948 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00586792_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053714.1 NP_446166.1 1 - 2 400 67
          GenBank mRNA AF393241.1 - 1 - 2 400 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000689.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankh

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphate ion transport
          biological_process
          regulation of bone mineralization
          inorganic diphosphate transport
          ion transmembrane transport
          phosphate ion transmembrane transport
          molecular_function
          inorganic phosphate transmembrane transporter activity
          phosphate ion transmembrane transporter activity
          inorganic diphosphate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P070522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152731 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115340 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58231 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1514 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146552 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070522 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070522 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P070522 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461470 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104839 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99851 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19577 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64114 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102978 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9584 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813970 - -
          Affymetrix Rat Gene 1.0 ST Array 10813974 - -
          Affymetrix Rat Gene 1.0 ST Array 10813979 - -
          Affymetrix Rat Gene 1.0 ST Array 10813973 - -
          Affymetrix Rat Gene 1.0 ST Array 10813977 - -
          Affymetrix Rat Gene 1.0 ST Array 10813975 - -
          Affymetrix Rat Gene 1.0 ST Array 10813981 - -
          Affymetrix Rat Gene 1.0 ST Array 10813972 - -
          Affymetrix Rat Gene 1.0 ST Array 10813980 - -
          Affymetrix Rat Gene 1.0 ST Array 10813976 - -
          Affymetrix Rat Gene 1.0 ST Array 10813971 - -
          Affymetrix Rat Gene 1.0 ST Array 10813978 - -
          Affymetrix Rat Gene 1.1 ST Array 10813969 - -
          Rat Genome 230 2.0 Array 1369249_PM_at - -
          Rat Genome 230 2.0 Array 1386344_at - -
          Rat Genome 230 2.0 Array 1393439_a_at - -
          Rat Genome 230 2.0 Array 1369249_at - -
          Rat Genome 230 2.0 Array 1393439_PM_a_at - -
          Rat Genome 230 2.0 Array 1386344_PM_at - -
          Affymetrix HT Rat Focus Array 1386344_PM_at - -
          Affymetrix HT Rat Focus Array 1393439_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369249_PM_at - -
          Affymetrix HT Rat Focus Array 1393439_a_at - -
          Affymetrix HT Rat Focus Array 1386344_at - -
          Affymetrix HT Rat Focus Array 1369249_at - -
          Affymetrix Rat Clariom S Assay PSR0200004164.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004167.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004169.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004168.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004161.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004165.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004166.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004170.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004162.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004160.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004163.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004171.rn.1 TC0200000689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200004172.rn.1 TC0200000689.rn.1 RTA TC
          Rat Expression Set 230 1369249_PM_at - -
          Rat Expression Set 230 1369249_at - -
          Rat Expression Set 230 1393439_a_at - -
          Rat Expression Set 230 1393439_PM_a_at - -
          Rat Expression Set 230 1386344_PM_at - -
          Rat Expression Set 230 1386344_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813969 - -
          Rat Genome U34 Set rc_AI045767_g_at - -
          Rat Genome U34 Set rc_AI045767_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21269846 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354148 - -
          RatRef-12 Expression BeadChip ILMN_48164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline