Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587074_m1
          See other SUCLG1 GE Assays ›
          Gene Symbol:
          Suclg1
          Gene Name:
          succinate-CoA ligase, alpha subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 101181302 - 101210692 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587074_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Suclg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053752.2 NP_446204.2 1 - 2 114 54
          XM_008762973.1 XP_008761195.1 1 - 2 144 54
          GenBank mRNA BC061537.1 - 1 - 2 118 6920009 54
          FQ212668.1 - 1 - 2 111 54
          FQ214532.1 - 1 - 2 73 54
          FQ214959.1 - 1 - 2 103 54
          FQ215028.1 - 1 - 2 72 54
          FQ215469.1 - 1 - 2 111 54
          FQ216893.1 - 1 - 2 103 54
          U75394.1 - 1 - 2 55 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001140.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Suclg1

          Panther Classification:

          Molecular Function -

          ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          succinyl-CoA metabolic process
          succinate metabolic process
          ATP citrate synthase activity
          succinate-CoA ligase (ADP-forming) activity
          succinate-CoA ligase (GDP-forming) activity
          GTP binding
          GDP binding
          poly(A) RNA binding
          protein heterodimerization activity
          cofactor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P399999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743752 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4743 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73883 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P399999 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P399999 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P399999 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P399999 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72800 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856438 - -
          Affymetrix Rat Gene 1.0 ST Array 10856437 - -
          Affymetrix Rat Gene 1.0 ST Array 10856440 - -
          Affymetrix Rat Gene 1.0 ST Array 10856442 - -
          Affymetrix Rat Gene 1.0 ST Array 10856444 - -
          Affymetrix Rat Gene 1.0 ST Array 10856445 - -
          Affymetrix Rat Gene 1.0 ST Array 10856441 - -
          Affymetrix Rat Gene 1.0 ST Array 10856439 - -
          Affymetrix Rat Gene 1.0 ST Array 10856443 - -
          Affymetrix Rat Gene 1.1 ST Array 10856436 - -
          Rat Genome 230 2.0 Array 1367642_PM_at - -
          Rat Genome 230 2.0 Array 1367642_at - -
          Affymetrix HT Rat Focus Array 1367642_PM_at - -
          Affymetrix HT Rat Focus Array 1367642_at - -
          Affymetrix Rat Clariom S Assay PSR0400008157.rn.1 TC0400001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008158.rn.1 TC0400001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008160.rn.1 TC0400001140.rn.1 RTA TC
          Rat Expression Set 230 1367642_at - -
          Rat Expression Set 230 1367642_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856436 - -
          Rat Genome U34 Set J03621_at - -
          Rat Genome U34 Set U75393_s_at - -
          Rat Genome U34 Set rc_AI104685_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22280993 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372406 - -
          RatRef-12 Expression BeadChip ILMN_56907 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline