Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00588808_m1
          See other MYO1B GE Assays ›
          Gene Symbol:
          Myo1b
          Gene Name:
          myosin Ib
          Gene Aliases:
          Myr1
          Chromosome Location:
          Chr.9: 54558202 - 54705350 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00588808_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myo1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053986.1 NP_446438.1 639 84
          GenBank mRNA X68199.1 - 639 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000628.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myo1b

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          post-Golgi vesicle-mediated transport
          actin filament organization
          nervous system development
          metabolic process
          actin filament-based movement
          actin filament bundle assembly
          microfilament motor activity
          motor activity
          actin binding
          calmodulin binding
          ATP binding
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          actin-dependent ATPase activity
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P016828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19724 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162447 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150111 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94368 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8213 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P016828 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557210 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701311721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69738 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76707 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104447 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39240 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43651 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5467 - -
          Agilent Rat cDNA Microarray (G4105A) 701311721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923258 - -
          Affymetrix Rat Gene 1.0 ST Array 10923259 - -
          Affymetrix Rat Gene 1.0 ST Array 10923243 - -
          Affymetrix Rat Gene 1.0 ST Array 10923255 - -
          Affymetrix Rat Gene 1.0 ST Array 10923241 - -
          Affymetrix Rat Gene 1.0 ST Array 10923248 - -
          Affymetrix Rat Gene 1.0 ST Array 10923254 - -
          Affymetrix Rat Gene 1.0 ST Array 10923263 - -
          Affymetrix Rat Gene 1.0 ST Array 10923246 - -
          Affymetrix Rat Gene 1.0 ST Array 10923249 - -
          Affymetrix Rat Gene 1.0 ST Array 10923266 - -
          Affymetrix Rat Gene 1.0 ST Array 10923247 - -
          Affymetrix Rat Gene 1.0 ST Array 10923256 - -
          Affymetrix Rat Gene 1.0 ST Array 10923242 - -
          Affymetrix Rat Gene 1.0 ST Array 10923252 - -
          Affymetrix Rat Gene 1.0 ST Array 10923238 - -
          Affymetrix Rat Gene 1.0 ST Array 10923244 - -
          Affymetrix Rat Gene 1.0 ST Array 10923264 - -
          Affymetrix Rat Gene 1.0 ST Array 10923257 - -
          Affymetrix Rat Gene 1.0 ST Array 10923240 - -
          Affymetrix Rat Gene 1.0 ST Array 10923253 - -
          Affymetrix Rat Gene 1.0 ST Array 10923269 - -
          Affymetrix Rat Gene 1.0 ST Array 10923265 - -
          Affymetrix Rat Gene 1.0 ST Array 10923260 - -
          Affymetrix Rat Gene 1.0 ST Array 10923251 - -
          Affymetrix Rat Gene 1.0 ST Array 10923262 - -
          Affymetrix Rat Gene 1.0 ST Array 10923268 - -
          Affymetrix Rat Gene 1.0 ST Array 10923267 - -
          Affymetrix Rat Gene 1.0 ST Array 10923261 - -
          Affymetrix Rat Gene 1.1 ST Array 10923237 - -
          Rat Genome 230 2.0 Array 1369720_PM_at - -
          Rat Genome 230 2.0 Array 1395485_s_at - -
          Rat Genome 230 2.0 Array 1395485_PM_s_at - -
          Rat Genome 230 2.0 Array 1397590_PM_at - -
          Rat Genome 230 2.0 Array 1397590_at - -
          Rat Genome 230 2.0 Array 1369720_at - -
          Affymetrix HT Rat Focus Array 1369720_PM_at - -
          Affymetrix HT Rat Focus Array 1397590_PM_at - -
          Affymetrix HT Rat Focus Array 1395485_s_at - -
          Affymetrix HT Rat Focus Array 1369720_at - -
          Affymetrix HT Rat Focus Array 1397590_at - -
          Affymetrix HT Rat Focus Array 1395485_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0900005086.rn.1 TC0900000628.rn.1 RTA TC
          Rat Expression Set 230 1369720_at - -
          Rat Expression Set 230 1369720_PM_at - -
          Rat Expression Set 230 1397590_at - -
          Rat Expression Set 230 1395485_s_at - -
          Rat Expression Set 230 1395485_PM_s_at - -
          Rat Expression Set 230 1397590_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923237 - -
          Rat Genome U34 Set X68199_at - -
          Rat Genome U34 Set rc_AA859008_at - -
          Rat Genome U34 Set X68199_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22393837 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373747 - -
          RatRef-12 Expression BeadChip ILMN_63519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline