Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589936_s1
          See other CHRM1 GE Assays ›
          Gene Symbol:
          Chrm1
          Gene Name:
          cholinergic receptor, muscarinic 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 224869087 - 224884475 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589936_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080773.1 NP_542951.1 1 950 77
          XM_006230974.2 XP_006231036.1 2 1643 77
          XM_008760215.1 XP_008758437.1 2 1462 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrm1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled acetylcholine receptor signaling pathway
          neuromuscular synaptic transmission
          regulation of locomotion
          positive regulation of ion transport
          saliva secretion
          cognition
          positive regulation of intracellular protein transport
          drug binding
          G-protein coupled acetylcholine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64927 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63043 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036499 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036499 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036499 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15593 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P545705 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 481 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75106 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713582 - -
          Affymetrix Rat Gene 1.1 ST Array 10713581 - -
          Rat Genome 230 2.0 Array 1387736_PM_at - -
          Rat Genome 230 2.0 Array 1387736_at - -
          Affymetrix HT Rat Focus Array 1387736_PM_at - -
          Affymetrix HT Rat Focus Array 1387736_at - -
          Rat Expression Set 230 1387736_PM_at - -
          Rat Expression Set 230 1387736_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713581 - -
          Rat Genome U34 Set AJ006522mRNA_at - -
          Rat Genome U34 Set M16406cds_s_at - -
          Rat Genome U34 Set M16406cds_s_at - -
          Rat Genome U34 Set AJ006522mRNA_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20853443 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361649 - -
          RatRef-12 Expression BeadChip ILMN_55978 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline