Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590432_m1
          See other LCP2 GE Assays ›
          Gene Symbol:
          Lcp2
          Gene Name:
          lymphocyte cytosolic protein 2
          Gene Aliases:
          Slp76
          Chromosome Location:
          Chr.10: 18996523 - 19043289 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590432_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130421.2 NP_569105.1 8 - 9 788 56
          GenBank mRNA AB072980.1 - 8 - 9 628 56
          BC100066.1 - 8 - 9 788 7387128 56
          FQ232596.1 - 8 - 9 802 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000338.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcp2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s)

          mast cell activation
          cytokine secretion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10190 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13117 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13117 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13117 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P864560 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13117 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104442 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103288 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732740 - -
          Affymetrix Rat Gene 1.0 ST Array 10732727 - -
          Affymetrix Rat Gene 1.0 ST Array 10732732 - -
          Affymetrix Rat Gene 1.0 ST Array 10732743 - -
          Affymetrix Rat Gene 1.0 ST Array 10732730 - -
          Affymetrix Rat Gene 1.0 ST Array 10732734 - -
          Affymetrix Rat Gene 1.0 ST Array 10732736 - -
          Affymetrix Rat Gene 1.0 ST Array 10732745 - -
          Affymetrix Rat Gene 1.0 ST Array 10732726 - -
          Affymetrix Rat Gene 1.0 ST Array 10732728 - -
          Affymetrix Rat Gene 1.0 ST Array 10732742 - -
          Affymetrix Rat Gene 1.0 ST Array 10732729 - -
          Affymetrix Rat Gene 1.0 ST Array 10732737 - -
          Affymetrix Rat Gene 1.0 ST Array 10732744 - -
          Affymetrix Rat Gene 1.0 ST Array 10732735 - -
          Affymetrix Rat Gene 1.0 ST Array 10732741 - -
          Affymetrix Rat Gene 1.0 ST Array 10732739 - -
          Affymetrix Rat Gene 1.0 ST Array 10732733 - -
          Affymetrix Rat Gene 1.0 ST Array 10732738 - -
          Affymetrix Rat Gene 1.0 ST Array 10732731 - -
          Affymetrix Rat Gene 1.1 ST Array 10732725 - -
          Rat Genome 230 2.0 Array 1390797_at - -
          Rat Genome 230 2.0 Array 1370090_at - -
          Rat Genome 230 2.0 Array 1370090_PM_at - -
          Rat Genome 230 2.0 Array 1390797_PM_at - -
          Affymetrix HT Rat Focus Array 1390797_at - -
          Affymetrix HT Rat Focus Array 1370090_at - -
          Affymetrix HT Rat Focus Array 1390797_PM_at - -
          Affymetrix HT Rat Focus Array 1370090_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000003232.rn.1 TC1000000338.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003233.rn.1 TC1000000338.rn.1 RTA TC
          Rat Expression Set 230 1370090_at - -
          Rat Expression Set 230 1370090_PM_at - -
          Rat Expression Set 230 1390797_PM_at - -
          Rat Expression Set 230 1390797_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732725 - -
          Rat Genome U34 Set rc_AA892316_at - -
          Rat Genome U34 Set rc_AI060017_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22125441 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365475 - -
          RatRef-12 Expression BeadChip ILMN_53609 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline