Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591030_m1
          See other CES1D GE Assays ›
          Gene Symbol:
          Ces1d
          Gene Name:
          carboxylesterase 1D
          Gene Aliases:
          Ces3
          Chromosome Location:
          Chr.19: 15195514 - 15239827 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591030_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces1d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133295.3 NP_579829.3 10 - 11 1215 67
          GenBank mRNA AF171640.1 - 10 - 11 1164 67
          BC061789.1 - 10 - 11 1215 5623732 67
          CF111142.1 - 7 - 8 715 67
          CO574231.1 - 6 - 7 537 67
          L46791.1 - 10 - 11 1194 67
          X51974.1 - 10 - 11 1212 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ces1d

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid catabolic process
          short-chain fatty acid catabolic process
          epithelial cell differentiation
          very-low-density lipoprotein particle assembly
          acylglycerol catabolic process
          sterol esterase activity
          triglyceride lipase activity
          fatty-acyl-ethyl-ester synthase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4265 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148413 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148413 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P570313 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700301086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61226 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28484 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62847 - -
          Agilent Rat cDNA Microarray (G4105A) 700301086 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806227 - -
          Affymetrix Rat Gene 1.0 ST Array 10931282 - -
          Affymetrix Rat Gene 1.0 ST Array 10931284 - -
          Affymetrix Rat Gene 1.0 ST Array 10806224 - -
          Affymetrix Rat Gene 1.0 ST Array 10806222 - -
          Affymetrix Rat Gene 1.0 ST Array 10931279 - -
          Affymetrix Rat Gene 1.0 ST Array 10931276 - -
          Affymetrix Rat Gene 1.0 ST Array 10931274 - -
          Affymetrix Rat Gene 1.0 ST Array 10931272 - -
          Affymetrix Rat Gene 1.0 ST Array 10806223 - -
          Affymetrix Rat Gene 1.0 ST Array 10806228 - -
          Affymetrix Rat Gene 1.0 ST Array 10931283 - -
          Affymetrix Rat Gene 1.0 ST Array 10931277 - -
          Affymetrix Rat Gene 1.0 ST Array 10931278 - -
          Affymetrix Rat Gene 1.0 ST Array 10806226 - -
          Affymetrix Rat Gene 1.0 ST Array 10806225 - -
          Affymetrix Rat Gene 1.0 ST Array 10931281 - -
          Affymetrix Rat Gene 1.0 ST Array 10931273 - -
          Affymetrix Rat Gene 1.1 ST Array 10931271 - -
          Affymetrix Rat Gene 1.1 ST Array 10806221 - -
          Rat Genome 230 2.0 Array 1370363_at - -
          Rat Genome 230 2.0 Array 1370363_PM_at - -
          Rat Genome 230 2.0 Array 1397427_at - -
          Rat Genome 230 2.0 Array 1397427_PM_at - -
          Affymetrix HT Rat Focus Array 1397427_at - -
          Affymetrix HT Rat Focus Array 1370363_at - -
          Affymetrix HT Rat Focus Array 1370363_PM_at - -
          Affymetrix HT Rat Focus Array 1397427_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900001248.rn.1 TC1900000150.rn.1 RTA TC
          Rat Expression Set 230 1370363_PM_at - -
          Rat Expression Set 230 1370363_at - -
          Rat Expression Set 230 1397427_PM_at - -
          Rat Expression Set 230 1397427_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806221 - -
          Affymetrix Rat Gene 1.0 ST Array 10931271 - -
          Rat Genome U34 Set L46791_at - -
          Rat Genome U34 Set rc_AA800851_r_at - -
          Rat Genome U34 Set rc_AA800851_i_at - -
          Rat Genome U34 Set X65296cds_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376983 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline