Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591465_m1
          See other AGPAT4 GE Assays ›
          Gene Symbol:
          Agpat4
          Gene Name:
          1-acylglycerol-3-phosphate O-acyltransferase 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 48719632 - 48826302 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591465_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Agpat4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133406.1 NP_596897.1 5 - 6 873 95
          XM_006227856.2 XP_006227918.1 5 - 6 1588 95
          XM_006227857.2 XP_006227919.1 5 - 6 1242 95
          GenBank mRNA AB067572.1 - 5 - 6 873 95
          BC086992.1 - 5 - 6 792 7191675 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004464.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Agpat4

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          CDP-diacylglycerol biosynthetic process
          1-acylglycerol-3-phosphate O-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142178 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165841 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13176 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13176 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13176 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13176 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103873 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28914 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7720 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66340 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68592 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74544 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718003 - -
          Affymetrix Rat Gene 1.0 ST Array 10718008 - -
          Affymetrix Rat Gene 1.0 ST Array 10718007 - -
          Affymetrix Rat Gene 1.0 ST Array 10718006 - -
          Affymetrix Rat Gene 1.0 ST Array 10718002 - -
          Affymetrix Rat Gene 1.0 ST Array 10718004 - -
          Affymetrix Rat Gene 1.0 ST Array 10718009 - -
          Affymetrix Rat Gene 1.0 ST Array 10718005 - -
          Affymetrix Rat Gene 1.1 ST Array 10718001 - -
          Rat Genome 230 2.0 Array 1368194_PM_at - -
          Rat Genome 230 2.0 Array 1368194_at - -
          Affymetrix HT Rat Focus Array 1368194_at - -
          Affymetrix HT Rat Focus Array 1368194_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100035481.rn.1 TC0100004464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100035482.rn.1 TC0100004464.rn.1 RTA TC
          Rat Expression Set 230 1368194_PM_at - -
          Rat Expression Set 230 1368194_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718001 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21397072 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372733 - -
          RatRef-12 Expression BeadChip ILMN_56708 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline