Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591831_g1
          See other NKG7 GE Assays ›
          Gene Symbol:
          Nkg7
          Gene Name:
          natural killer cell granule protein 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 98492873 - 98493940 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591831_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkg7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133540.1 NP_598224.1 2 - 3 447 63
          GenBank mRNA AF082535.1 - 2 - 3 447 63
          FQ222255.1 - 2 - 3 453 63
          FQ226323.1 - 2 - 3 453 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005306.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nkg7

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P500080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8916 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P500080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P500080 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P500080 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P500080 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700542767 - -
          Agilent Rat cDNA Microarray (G4105A) 700542767 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706332 - -
          Affymetrix Rat Gene 1.0 ST Array 10706331 - -
          Affymetrix Rat Gene 1.0 ST Array 10706328 - -
          Affymetrix Rat Gene 1.0 ST Array 10706330 - -
          Affymetrix Rat Gene 1.0 ST Array 10706329 - -
          Affymetrix Rat Gene 1.0 ST Array 10706327 - -
          Affymetrix Rat Gene 1.1 ST Array 10706326 - -
          Rat Genome 230 2.0 Array 1368455_PM_at - -
          Rat Genome 230 2.0 Array 1368455_at - -
          Affymetrix HT Rat Focus Array 1368455_at - -
          Affymetrix HT Rat Focus Array 1368455_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100042813.rn.1 TC0100005306.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042812.rn.1 TC0100005306.rn.1 RTA TC
          Rat Expression Set 230 1368455_at - -
          Rat Expression Set 230 1368455_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706326 - -
          Rat Genome U34 Set rc_AI178608_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21799163 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360097 - -
          RatRef-12 Expression BeadChip ILMN_62827 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline