Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592200_m1
          See other OPA1 GE Assays ›
          Gene Symbol:
          Opa1
          Gene Name:
          optic atrophy 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 74717600 - 74793902 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592200_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Opa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133585.3 NP_598269.3 23 - 24 2486 132
          XM_006248497.1 XP_006248559.1 25 - 26 2750 132
          XM_006248498.1 XP_006248560.1 24 - 25 2696 132
          XM_006248499.1 XP_006248561.1 24 - 25 2639 132
          GenBank mRNA AY333988.1 - 23 - 24 2395 132
          AY510274.1 - 23 - 24 2541 132
          BC111071.1 - 23 - 24 2486 7324017 132
          U93197.1 - 7 - 8 692 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001821.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Opa1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          neural tube closure
          apoptotic process
          mitochondrion organization
          inner mitochondrial membrane organization
          aging
          visual perception
          mitochondrial fusion
          metabolic process
          positive regulation of mitochondrial fusion
          positive regulation of neuron maturation
          negative regulation of release of cytochrome c from mitochondria
          cellular senescence
          positive regulation of dendrite development
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of intrinsic apoptotic signaling pathway
          molecular_function
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P409729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139156 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36133 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409729 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409729 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P409729 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409729 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P427200 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701621750 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97131 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19575 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34722 - -
          Agilent Rat cDNA Microarray (G4105A) 701621750 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754972 - -
          Affymetrix Rat Gene 1.0 ST Array 10754965 - -
          Affymetrix Rat Gene 1.0 ST Array 10754966 - -
          Affymetrix Rat Gene 1.0 ST Array 10754979 - -
          Affymetrix Rat Gene 1.0 ST Array 10754974 - -
          Affymetrix Rat Gene 1.0 ST Array 10754961 - -
          Affymetrix Rat Gene 1.0 ST Array 10754952 - -
          Affymetrix Rat Gene 1.0 ST Array 10754970 - -
          Affymetrix Rat Gene 1.0 ST Array 10754955 - -
          Affymetrix Rat Gene 1.0 ST Array 10754956 - -
          Affymetrix Rat Gene 1.0 ST Array 10754957 - -
          Affymetrix Rat Gene 1.0 ST Array 10754980 - -
          Affymetrix Rat Gene 1.0 ST Array 10754982 - -
          Affymetrix Rat Gene 1.0 ST Array 10754981 - -
          Affymetrix Rat Gene 1.0 ST Array 10754960 - -
          Affymetrix Rat Gene 1.0 ST Array 10754971 - -
          Affymetrix Rat Gene 1.0 ST Array 10754959 - -
          Affymetrix Rat Gene 1.0 ST Array 10754978 - -
          Affymetrix Rat Gene 1.0 ST Array 10754969 - -
          Affymetrix Rat Gene 1.0 ST Array 10754953 - -
          Affymetrix Rat Gene 1.0 ST Array 10754976 - -
          Affymetrix Rat Gene 1.0 ST Array 10754951 - -
          Affymetrix Rat Gene 1.0 ST Array 10754967 - -
          Affymetrix Rat Gene 1.0 ST Array 10754977 - -
          Affymetrix Rat Gene 1.0 ST Array 10754954 - -
          Affymetrix Rat Gene 1.0 ST Array 10754962 - -
          Affymetrix Rat Gene 1.0 ST Array 10754958 - -
          Affymetrix Rat Gene 1.0 ST Array 10754964 - -
          Affymetrix Rat Gene 1.0 ST Array 10754963 - -
          Affymetrix Rat Gene 1.0 ST Array 10754968 - -
          Affymetrix Rat Gene 1.1 ST Array 10754950 - -
          Rat Genome 230 2.0 Array 1387225_at - -
          Rat Genome 230 2.0 Array 1387225_PM_at - -
          Affymetrix HT Rat Focus Array 1387225_at - -
          Affymetrix HT Rat Focus Array 1387225_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100011548.rn.1 TC1100001821.rn.1 RTA TC
          Rat Expression Set 230 1387225_at - -
          Rat Expression Set 230 1387225_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754950 - -
          Rat Genome U34 Set U93197_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21824950 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355429 - -
          RatRef-12 Expression BeadChip ILMN_62200 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline