Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593441_m1
          See other PRPF18 GE Assays ›
          Gene Symbol:
          Prpf18
          Gene Name:
          pre-mRNA processing factor 18
          Gene Aliases:
          Pprf18
          Chromosome Location:
          Chr.17: 77601914 - 77632507 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593441_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prpf18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138523.2 NP_612532.1 5 - 6 614 82
          GenBank mRNA AF244920.1 - 5 - 6 609 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001008.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prpf18

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          potassium channel inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P491929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421097 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169045 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491929 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491929 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P491929 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491929 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702134670 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11827 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55378 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85815 - -
          Agilent Rat cDNA Microarray (G4105A) 702134670 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796382 - -
          Affymetrix Rat Gene 1.0 ST Array 10796377 - -
          Affymetrix Rat Gene 1.0 ST Array 10796372 - -
          Affymetrix Rat Gene 1.0 ST Array 10796370 - -
          Affymetrix Rat Gene 1.0 ST Array 10796380 - -
          Affymetrix Rat Gene 1.0 ST Array 10796367 - -
          Affymetrix Rat Gene 1.0 ST Array 10796374 - -
          Affymetrix Rat Gene 1.0 ST Array 10796379 - -
          Affymetrix Rat Gene 1.0 ST Array 10796369 - -
          Affymetrix Rat Gene 1.0 ST Array 10796375 - -
          Affymetrix Rat Gene 1.0 ST Array 10796376 - -
          Affymetrix Rat Gene 1.0 ST Array 10796371 - -
          Affymetrix Rat Gene 1.0 ST Array 10796378 - -
          Affymetrix Rat Gene 1.0 ST Array 10796368 - -
          Affymetrix Rat Gene 1.0 ST Array 10796373 - -
          Affymetrix Rat Gene 1.1 ST Array 10796366 - -
          Affymetrix Rat Gene 1.1 ST Array 10796381 - -
          Rat Genome 230 2.0 Array 1388005_at - -
          Rat Genome 230 2.0 Array 1399021_PM_at - -
          Rat Genome 230 2.0 Array 1388005_PM_at - -
          Rat Genome 230 2.0 Array 1399021_at - -
          Affymetrix HT Rat Focus Array 1388005_at - -
          Affymetrix HT Rat Focus Array 1399021_PM_at - -
          Affymetrix HT Rat Focus Array 1399021_at - -
          Affymetrix HT Rat Focus Array 1388005_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700006945.rn.1 TC1700001008.rn.1 RTA TC
          Rat Expression Set 230 1388005_PM_at - -
          Rat Expression Set 230 1388005_at - -
          Rat Expression Set 230 1399021_PM_at - -
          Rat Expression Set 230 1399021_at - -
          Affymetrix Rat Gene 1.0 ST Array 10796366 - -
          Affymetrix Rat Gene 1.0 ST Array 10796381 - -
          Rat Genome U34 Set rc_AI029421_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21358772 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353121 - -
          RatRef-12 Expression BeadChip ILMN_50750 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline