Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593485_m1
          See other PBLD1 GE Assays ›
          Gene Symbol:
          Pbld1
          Gene Name:
          phenazine biosynthesis-like protein domain containing 1
          Gene Aliases:
          Mawbp, Pbld
          Chromosome Location:
          Chr.20: 27103300 - 27118652 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593485_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pbld1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138530.2 NP_612539.2 424 95
          XM_006256372.2 XP_006256434.1 6 - 7 739 95
          GenBank mRNA AY083160.1 - 436 95
          BC078735.1 - 424 7126368 95
          FQ209640.1 - 459 95
          FQ219037.1 - 459 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pbld1

          Gene Ontology Categories:

          Function(s) Process(es)

          biosynthetic process
          negative regulation of epithelial cell migration
          negative regulation of epithelial to mesenchymal transition
          maintenance of gastrointestinal epithelium
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of epithelial cell proliferation
          negative regulation of SMAD protein import into nucleus
          negative regulation of pathway-restricted SMAD protein phosphorylation
          isomerase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13290 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13290 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13290 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73021 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829894 - -
          Affymetrix Rat Gene 1.0 ST Array 10829892 - -
          Affymetrix Rat Gene 1.0 ST Array 10829890 - -
          Affymetrix Rat Gene 1.0 ST Array 10829889 - -
          Affymetrix Rat Gene 1.0 ST Array 10829891 - -
          Affymetrix Rat Gene 1.0 ST Array 10829895 - -
          Affymetrix Rat Gene 1.0 ST Array 10829893 - -
          Affymetrix Rat Gene 1.1 ST Array 10829888 - -
          Rat Genome 230 2.0 Array 1370320_PM_at - -
          Rat Genome 230 2.0 Array 1390993_at - -
          Rat Genome 230 2.0 Array 1370320_at - -
          Rat Genome 230 2.0 Array 1390993_PM_at - -
          Affymetrix HT Rat Focus Array 1370320_at - -
          Affymetrix HT Rat Focus Array 1390993_PM_at - -
          Affymetrix HT Rat Focus Array 1390993_at - -
          Affymetrix HT Rat Focus Array 1370320_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000012036.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012040.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012043.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012035.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012046.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012044.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012038.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012048.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012039.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012042.rn.1 TC2000001477.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012045.rn.1 TC2000001477.rn.1 RTA TC
          Rat Expression Set 230 1370320_PM_at - -
          Rat Expression Set 230 1370320_at - -
          Rat Expression Set 230 1390993_at - -
          Rat Expression Set 230 1390993_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829888 - -
          Rat Genome U34 Set rc_AI233773_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21153550 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371795 - -
          RatRef-12 Expression BeadChip ILMN_66922 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline