Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593631_g1
          See other TECR GE Assays ›
          Gene Symbol:
          Tecr
          Gene Name:
          trans-2,3-enoyl-CoA reductase
          Gene Aliases:
          Gpsn2, SC2
          Chromosome Location:
          Chr.19: 24705478 - 24732029 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593631_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tecr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138549.1 NP_612558.1 11 - 12 895 79
          XM_006255216.2 XP_006255278.1 10 - 11 1030 79
          XM_006255217.2 XP_006255279.1 9 - 10 897 79
          XM_006255218.2 XP_006255280.1 10 - 11 855 79
          GenBank mRNA BC059115.1 - 11 - 12 858 6920522 79
          EU000473.1 - 11 - 12 858 79
          FQ215654.1 - 10 - 11 952 79
          FQ215778.1 - 9 - 10 735 79
          FQ216428.1 - 9 - 10 902 79
          FQ220726.1 - 11 - 12 844 79
          S45663.1 - 11 - 12 895 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tecr

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          steroid biosynthetic process
          fatty acid elongation
          very long-chain fatty acid biosynthetic process
          oxidation-reduction process
          oxidoreductase activity, acting on the CH-CH group of donors

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P021636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148728 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021636 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021636 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P021636 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13294 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1053105 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700291633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9112 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76309 - -
          Agilent Rat cDNA Microarray (G4105A) 700291633 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806859 - -
          Affymetrix Rat Gene 1.0 ST Array 10806862 - -
          Affymetrix Rat Gene 1.0 ST Array 10806856 - -
          Affymetrix Rat Gene 1.0 ST Array 10806860 - -
          Affymetrix Rat Gene 1.0 ST Array 10806861 - -
          Affymetrix Rat Gene 1.0 ST Array 10806863 - -
          Affymetrix Rat Gene 1.0 ST Array 10806858 - -
          Affymetrix Rat Gene 1.0 ST Array 10717934 - -
          Affymetrix Rat Gene 1.1 ST Array 10806855 - -
          Affymetrix Rat Gene 1.1 ST Array 10717933 - -
          Rat Genome 230 2.0 Array 1371358_at - -
          Rat Genome 230 2.0 Array 1371358_PM_at - -
          Affymetrix HT Rat Focus Array 1371358_PM_at - -
          Affymetrix HT Rat Focus Array 1371358_at - -
          Affymetrix Rat Clariom S Assay PSR1900009361.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009351.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009352.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009359.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009353.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009365.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009349.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009354.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009360.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009371.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009357.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009364.rn.1 TC1900001172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900009373.rn.1 TC1900001172.rn.1 RTA TC
          Rat Expression Set 230 1371358_PM_at - -
          Rat Expression Set 230 1371358_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806855 - -
          Affymetrix Rat Gene 1.0 ST Array 10717933 - -
          Rat Genome U34 Set S45663_g_at - -
          Rat Genome U34 Set S45663_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21929699 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366848 - -
          RatRef-12 Expression BeadChip ILMN_58885 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline