Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593654_m1
          See other DAB2IP GE Assays ›
          Gene Symbol:
          Dab2ip
          Gene Name:
          DAB2 interacting protein
          Gene Aliases:
          AIP-1, DIP1/2
          Chromosome Location:
          Chr.3: 14889263 - 15060286 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593654_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dab2ip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138710.3 NP_619724.3 14 - 15 3479 63
          XM_006234057.2 XP_006234119.1 13 - 14 3466 63
          XM_008761697.1 XP_008759919.1 11 - 12 3877 63
          XM_008761698.1 XP_008759920.1 11 - 12 3811 63
          XM_008761699.1 XP_008759921.1 11 - 12 3878 63
          GenBank mRNA AF236130.1 - 13 - 14 3187 63
          BC061865.1 - 10 - 11 2793 5598046 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000309.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dab2ip

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPKKK activity
          angiogenesis
          negative regulation of protein phosphorylation
          apoptotic process
          response to unfolded protein
          I-kappaB phosphorylation
          activation of JUN kinase activity
          negative regulation of cell proliferation
          extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell migration
          negative regulation of epithelial cell migration
          negative regulation of epithelial to mesenchymal transition
          positive regulation of neuron projection development
          negative regulation of phosphatidylinositol 3-kinase signaling
          negative regulation of angiogenesis
          cell motility involved in cerebral cortex radial glia guided migration
          layer formation in cerebral cortex
          negative regulation of cell growth
          negative regulation of vascular endothelial growth factor receptor signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of toll-like receptor 4 signaling pathway
          negative regulation of GTPase activity
          tube formation
          negative regulation of catenin import into nucleus
          cellular response to vascular endothelial growth factor stimulus
          vascular endothelial growth factor receptor-2 signaling pathway
          reelin-mediated signaling pathway
          negative regulation of epidermal growth factor receptor signaling pathway
          positive regulation of apoptotic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of protein complex assembly
          negative regulation of MAP kinase activity
          positive regulation of MAPK cascade
          regulation of protein heterodimerization activity
          positive regulation of JUN kinase activity
          negative regulation of phosphatidylinositol 3-kinase activity
          cellular protein catabolic process
          positive regulation of protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          negative regulation of Ras protein signal transduction
          negative regulation of fibroblast proliferation
          neuron projection morphogenesis
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          negative regulation of G0 to G1 transition
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of cell cycle arrest
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          negative regulation of protein serine/threonine kinase activity
          positive regulation of protein serine/threonine kinase activity
          negative regulation of canonical Wnt signaling pathway
          positive regulation of synapse maturation
          positive regulation of dendrite development
          regulation of p38MAPK cascade
          negative regulation of vascular endothelial growth factor signaling pathway
          positive regulation of proteasomal protein catabolic process
          negative regulation of cyclin catabolic process
          positive regulation of neuron migration
          positive regulation of apoptotic signaling pathway
          GTPase activator activity
          death receptor binding
          protein binding
          SH3 domain binding
          kinase binding
          protein kinase binding
          mitogen-activated protein kinase kinase binding
          mitogen-activated protein kinase kinase kinase binding
          phosphatidylinositol-3-phosphate binding
          protein complex binding
          signaling adaptor activity
          Toll-like receptor 4 binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          identical protein binding
          protein homodimerization activity
          vascular endothelial growth factor receptor 2 binding
          phosphatidylinositol 3-kinase binding
          protein N-terminus binding
          protein phosphatase 2A binding
          phosphatidylinositol-4-phosphate binding
          14-3-3 protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69117 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74014 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72637 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124737 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107492 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123266 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123266 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372544 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P249161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56089 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41692 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23038 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39011 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835788 - -
          Affymetrix Rat Gene 1.0 ST Array 10835784 - -
          Affymetrix Rat Gene 1.0 ST Array 10835786 - -
          Affymetrix Rat Gene 1.0 ST Array 10835792 - -
          Affymetrix Rat Gene 1.0 ST Array 10835795 - -
          Affymetrix Rat Gene 1.0 ST Array 10835782 - -
          Affymetrix Rat Gene 1.0 ST Array 10835794 - -
          Affymetrix Rat Gene 1.0 ST Array 10835785 - -
          Affymetrix Rat Gene 1.0 ST Array 10835783 - -
          Affymetrix Rat Gene 1.0 ST Array 10835790 - -
          Affymetrix Rat Gene 1.0 ST Array 10835793 - -
          Affymetrix Rat Gene 1.0 ST Array 10835797 - -
          Affymetrix Rat Gene 1.0 ST Array 10835789 - -
          Affymetrix Rat Gene 1.0 ST Array 10835791 - -
          Affymetrix Rat Gene 1.0 ST Array 10835787 - -
          Affymetrix Rat Gene 1.0 ST Array 10835781 - -
          Affymetrix Rat Gene 1.0 ST Array 10835796 - -
          Affymetrix Rat Gene 1.1 ST Array 10835779 - -
          Rat Genome 230 2.0 Array 1370335_PM_at - -
          Rat Genome 230 2.0 Array 1370335_at - -
          Affymetrix HT Rat Focus Array 1370335_PM_at - -
          Affymetrix HT Rat Focus Array 1370335_at - -
          Affymetrix Rat Clariom S Assay PSR0300003464.rn.1 TC0300000309.rn.1 RTA TC
          Rat Expression Set 230 1370335_at - -
          Rat Expression Set 230 1370335_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835779 - -
          Rat Genome U34 Set rc_AI009629_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21393713 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373399 - -
          RatRef-12 Expression BeadChip ILMN_54865 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline