Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594010_m1
          See other TDRD7 GE Assays ›
          Gene Symbol:
          Tdrd7
          Gene Name:
          tudor domain containing 7
          Gene Aliases:
          Pctaire2bp
          Chromosome Location:
          Chr.5: 61557909 - 61631719 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594010_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tdrd7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138871.1 NP_620226.1 14 - 15 2628 84
          XM_006238107.2 XP_006238169.1 14 - 15 2459 84
          XM_006238108.2 XP_006238170.1 13 - 14 2392 84
          XM_006238111.2 XP_006238173.1 13 - 14 2629 84
          XM_008763704.1 XP_008761926.1 13 - 14 2786 84
          GenBank mRNA AB030644.1 - 14 - 15 2628 84
          BC090066.1 - 13 - 14 2347 7122915 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000580.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tdrd7

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lens morphogenesis in camera-type eye
          germ cell development
          spermatogenesis
          posttranscriptional regulation of gene expression
          lens fiber cell differentiation
          mRNA binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177853 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089987 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089987 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089987 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P264381 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700541074 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701094036 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57469 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40348 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43506 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80224 - -
          Agilent Rat cDNA Microarray (G4105A) 700541074 - -
          Agilent Rat cDNA Microarray (G4105A) 701094036 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868800 - -
          Affymetrix Rat Gene 1.0 ST Array 10868806 - -
          Affymetrix Rat Gene 1.0 ST Array 10868813 - -
          Affymetrix Rat Gene 1.0 ST Array 10868804 - -
          Affymetrix Rat Gene 1.0 ST Array 10868815 - -
          Affymetrix Rat Gene 1.0 ST Array 10868814 - -
          Affymetrix Rat Gene 1.0 ST Array 10868808 - -
          Affymetrix Rat Gene 1.0 ST Array 10868816 - -
          Affymetrix Rat Gene 1.0 ST Array 10868807 - -
          Affymetrix Rat Gene 1.0 ST Array 10868812 - -
          Affymetrix Rat Gene 1.0 ST Array 10868809 - -
          Affymetrix Rat Gene 1.0 ST Array 10868805 - -
          Affymetrix Rat Gene 1.0 ST Array 10868803 - -
          Affymetrix Rat Gene 1.0 ST Array 10868799 - -
          Affymetrix Rat Gene 1.0 ST Array 10868802 - -
          Affymetrix Rat Gene 1.0 ST Array 10868810 - -
          Affymetrix Rat Gene 1.0 ST Array 10868811 - -
          Affymetrix Rat Gene 1.0 ST Array 10868801 - -
          Affymetrix Rat Gene 1.1 ST Array 10868798 - -
          Rat Genome 230 2.0 Array 1370351_PM_at - -
          Rat Genome 230 2.0 Array 1385848_s_at - -
          Rat Genome 230 2.0 Array 1370351_at - -
          Rat Genome 230 2.0 Array 1385848_PM_s_at - -
          Affymetrix HT Rat Focus Array 1385848_s_at - -
          Affymetrix HT Rat Focus Array 1370351_PM_at - -
          Affymetrix HT Rat Focus Array 1385848_PM_s_at - -
          Affymetrix HT Rat Focus Array 1370351_at - -
          Affymetrix Rat Clariom S Assay PSR0500003858.rn.1 TC0500000580.rn.1 RTA TC
          Rat Expression Set 230 1370351_PM_at - -
          Rat Expression Set 230 1370351_at - -
          Rat Expression Set 230 1385848_PM_s_at - -
          Rat Expression Set 230 1385848_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868798 - -
          Rat Genome U34 Set rc_AA850501_at - -
          Rat Genome U34 Set rc_AI112175_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22368280 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350790 - -
          RatRef-12 Expression BeadChip ILMN_53021 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline