Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594422_m1
          See other FBXO6 GE Assays ›
          Gene Symbol:
          Fbxo6
          Gene Name:
          F-box protein 6
          Gene Aliases:
          Fbg2, Fbxo6b
          Chromosome Location:
          Chr.5: 164956313 - 164962072 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594422_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138917.3 NP_620272.3 1 - 2 280 110
          XM_006239372.2 XP_006239434.1 2 - 3 584 110
          XM_006239373.2 XP_006239435.1 2 - 3 517 110
          XM_006239374.2 XP_006239436.1 2 - 3 444 110
          XM_006239375.2 XP_006239437.1 2 - 3 448 110
          GenBank mRNA AF393484.1 - 1 - 2 280 110
          BC063148.1 - 1 - 2 282 6921385 110
          FQ232698.1 - 2 - 3 350 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004043.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo6

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          glycoprotein catabolic process
          response to unfolded protein
          protein ubiquitination
          ER-associated ubiquitin-dependent protein catabolic process
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          glycoprotein binding
          ubiquitin-protein transferase activity
          carbohydrate binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P402668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P402668 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P402668 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P402668 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402668 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48830 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881584 - -
          Affymetrix Rat Gene 1.0 ST Array 10881587 - -
          Affymetrix Rat Gene 1.0 ST Array 10881585 - -
          Affymetrix Rat Gene 1.0 ST Array 10881586 - -
          Affymetrix Rat Gene 1.0 ST Array 10881589 - -
          Affymetrix Rat Gene 1.0 ST Array 10881588 - -
          Affymetrix Rat Gene 1.1 ST Array 10881583 - -
          Rat Genome 230 2.0 Array 1370820_at - -
          Rat Genome 230 2.0 Array 1370820_PM_at - -
          Affymetrix HT Rat Focus Array 1370820_PM_at - -
          Affymetrix HT Rat Focus Array 1370820_at - -
          Affymetrix Rat Clariom S Assay PSR0500030797.rn.1 TC0500004043.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500030798.rn.1 TC0500004043.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500030799.rn.1 TC0500004043.rn.1 RTA TC
          Rat Expression Set 230 1370820_PM_at - -
          Rat Expression Set 230 1370820_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881583 - -
          Rat Genome U34 Set rc_AA848404_at - -
          Rat Genome U34 Set rc_AI071480_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21300130 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359050 - -
          RatRef-12 Expression BeadChip ILMN_52079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline