Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595146_m1
          See other SLC15A3 GE Assays ›
          Gene Symbol:
          Slc15a3
          Gene Name:
          solute carrier family 15 (oligopeptide transporter), member 3
          Gene Aliases:
          Pht2
          Chromosome Location:
          Chr.1: 226897853 - 226912644 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595146_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc15a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139341.1 NP_647557.1 2 - 3 932 120
          GenBank mRNA AB026665.1 - 2 - 3 932 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003210.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc15a3

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          protein catabolic process
          oligopeptide transmembrane transport
          proton-dependent oligopeptide secondary active transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P008091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008091 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008091 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P008091 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P270468 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53793 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713977 - -
          Affymetrix Rat Gene 1.0 ST Array 10713975 - -
          Affymetrix Rat Gene 1.0 ST Array 10713980 - -
          Affymetrix Rat Gene 1.0 ST Array 10713978 - -
          Affymetrix Rat Gene 1.0 ST Array 10713981 - -
          Affymetrix Rat Gene 1.0 ST Array 10713979 - -
          Affymetrix Rat Gene 1.0 ST Array 10713976 - -
          Affymetrix Rat Gene 1.0 ST Array 10713982 - -
          Affymetrix Rat Gene 1.1 ST Array 10713974 - -
          Rat Genome 230 2.0 Array 1370516_at - -
          Rat Genome 230 2.0 Array 1370516_PM_at - -
          Affymetrix HT Rat Focus Array 1370516_at - -
          Affymetrix HT Rat Focus Array 1370516_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100026555.rn.1 TC0100003210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026554.rn.1 TC0100003210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026556.rn.1 TC0100003210.rn.1 RTA TC
          Rat Expression Set 230 1370516_at - -
          Rat Expression Set 230 1370516_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713974 - -
          Rat Genome U34 Set rc_AI172497_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21356364 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349041 - -
          RatRef-12 Expression BeadChip ILMN_61391 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline