Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597326_m1
          See other ZC3HAV1 GE Assays ›
          Gene Symbol:
          Zc3hav1
          Gene Name:
          zinc finger CCCH type, antiviral 1
          Gene Aliases:
          ARTD13, Zap
          Chromosome Location:
          Chr.4: 66012111 - 66062156 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597326_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zc3hav1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173045.2 NP_766633.2 7 - 8 2417 112
          XM_006236295.2 XP_006236357.1 7 - 8 2478 112
          GenBank mRNA AF521008.1 - 7 - 8 2101 112
          BC131842.1 - 7 - 8 2417 8372019 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002711.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zc3hav1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to virus
          positive regulation of type I interferon production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          suppression by virus of host molecular function
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of viral genome replication
          innate immune response
          regulation of defense response to virus by host
          defense response to virus
          positive regulation of mRNA catabolic process
          cellular response to exogenous dsRNA
          positive regulation of RIG-I signaling pathway
          RNA binding
          protein binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P271423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129085 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31085 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271423 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271423 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P271423 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P271423 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85989 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10861977 - -
          Affymetrix Rat Gene 1.0 ST Array 10861979 - -
          Affymetrix Rat Gene 1.0 ST Array 10861984 - -
          Affymetrix Rat Gene 1.0 ST Array 10861982 - -
          Affymetrix Rat Gene 1.0 ST Array 10861980 - -
          Affymetrix Rat Gene 1.0 ST Array 10861978 - -
          Affymetrix Rat Gene 1.0 ST Array 10861983 - -
          Affymetrix Rat Gene 1.0 ST Array 10861981 - -
          Affymetrix Rat Gene 1.0 ST Array 10861985 - -
          Affymetrix Rat Gene 1.1 ST Array 10861976 - -
          Affymetrix Rat Clariom S Assay PSR0400019298.rn.1 TC0400002711.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10861976 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22039186 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356609 - -
          RatRef-12 Expression BeadChip ILMN_60588 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline