Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597468_m1
          See other BRINP2 GE Assays ›
          Gene Symbol:
          Brinp2
          Gene Name:
          bone morphogenetic protein/retinoic acid inducible neural-specific 2
          Gene Aliases:
          Fam5b
          Chromosome Location:
          Chr.13: 75948679 - 76049363 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597468_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brinp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173115.1 NP_775138.1 2 - 3 1152 98
          GenBank mRNA AB077853.1 - 2 - 3 1152 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001716.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Brinp2

          Gene Ontology Categories:

          Function(s)

          cell cycle arrest
          nervous system development
          positive regulation of neuron differentiation
          negative regulation of cell cycle
          negative regulation of mitotic cell cycle
          cellular response to retinoic acid

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143251 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1005082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131005 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110278 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129348 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167384 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130783 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1005082 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143251 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143251 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1005082 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1005082 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143251 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1005082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55861 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95202 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91590 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24002 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12081 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769077 - -
          Affymetrix Rat Gene 1.0 ST Array 10769084 - -
          Affymetrix Rat Gene 1.0 ST Array 10769079 - -
          Affymetrix Rat Gene 1.0 ST Array 10769080 - -
          Affymetrix Rat Gene 1.0 ST Array 10769081 - -
          Affymetrix Rat Gene 1.0 ST Array 10769082 - -
          Affymetrix Rat Gene 1.0 ST Array 10769083 - -
          Affymetrix Rat Gene 1.0 ST Array 10769078 - -
          Affymetrix Rat Gene 1.0 ST Array 10769076 - -
          Affymetrix Rat Gene 1.0 ST Array 10769085 - -
          Affymetrix Rat Gene 1.1 ST Array 10769075 - -
          Rat Genome 230 2.0 Array 1383370_PM_at - -
          Rat Genome 230 2.0 Array 1386774_PM_at - -
          Rat Genome 230 2.0 Array 1383370_at - -
          Rat Genome 230 2.0 Array 1386774_at - -
          Affymetrix HT Rat Focus Array 1386774_PM_at - -
          Affymetrix HT Rat Focus Array 1386774_at - -
          Affymetrix HT Rat Focus Array 1383370_at - -
          Affymetrix HT Rat Focus Array 1383370_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300012013.rn.1 TC1300001716.rn.1 RTA TC
          Rat Expression Set 230 1383370_at - -
          Rat Expression Set 230 1386774_at - -
          Rat Expression Set 230 1386774_PM_at - -
          Rat Expression Set 230 1383370_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769075 - -
          Rat Genome U34 Set rc_AI070394_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20965838 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351220 - -
          RatRef-12 Expression BeadChip ILMN_50326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline