Search
Search
| Gene Symbol: Mvp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022715.2 | NP_073206.2 | 3 - 4 | 380 | 106 | |
| GenBank mRNA | BC071174.1 | - | 3 - 4 | 387 | 7099451 | 106 |
| U09870.2 | - | 3 - 4 | 380 | 106 | ||
Set Membership: |
|