Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024143.2 | NP_077057.1 | 8 - 9 | 1813 | 93 | |
GenBank mRNA | AF242189.1 | - | 8 - 9 | 1814 | 93 | |
BC091147.1 | - | 8 - 9 | 1836 | 7370589 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |