Search
Search
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024157.1 | NP_077071.1 | 5 - 6 | 793 | 79 | |
| XM_008761526.1 | XP_008759748.1 | 5 - 6 | 942 | 79 | ||
| GenBank mRNA | BC089798.1 | - | 5 - 6 | 798 | 7321559 | 79 |
| Y18965.1 | - | 5 - 6 | 793 | 79 | ||
Set Membership: |
|