Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00666995_m1
          See other NR4A1 GE Assays ›
          Gene Symbol:
          Nr4a1
          Gene Name:
          nuclear receptor subfamily 4, group A, member 1
          Gene Aliases:
          HMR, Ngfi-b, Nur77
          Chromosome Location:
          Chr.7: 142900807 - 142920216 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00666995_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr4a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024388.2 NP_077364.2 6 - 7 1658 98
          XM_006242356.2 XP_006242418.1 6 - 7 4485 98
          XM_006242358.2 XP_006242420.1 6 - 7 2413 98
          GenBank mRNA BC097313.1 - 6 - 7 1651 7458540 98
          U17254.1 - 6 - 7 1651 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001870.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr4a1

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          cell migration involved in sprouting angiogenesis
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          intracellular receptor signaling pathway
          endothelial cell chemotaxis
          skeletal muscle cell differentiation
          cellular response to vascular endothelial growth factor stimulus
          positive regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          steroid hormone mediated signaling pathway
          cellular response to fibroblast growth factor stimulus
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cellular response to organic substance
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          zinc ion binding
          protein homodimerization activity
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P682589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P682589 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P682589 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P682589 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P682589 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700693437 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31911 - -
          Agilent Rat cDNA Microarray (G4105A) 700693437 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899389 - -
          Affymetrix Rat Gene 1.0 ST Array 10899394 - -
          Affymetrix Rat Gene 1.0 ST Array 10899392 - -
          Affymetrix Rat Gene 1.0 ST Array 10899395 - -
          Affymetrix Rat Gene 1.0 ST Array 10899393 - -
          Affymetrix Rat Gene 1.0 ST Array 10899391 - -
          Affymetrix Rat Gene 1.0 ST Array 10899388 - -
          Affymetrix Rat Gene 1.1 ST Array 10899387 - -
          Rat Genome 230 2.0 Array 1386935_at - -
          Rat Genome 230 2.0 Array 1386935_PM_at - -
          Affymetrix HT Rat Focus Array 1386935_at - -
          Affymetrix HT Rat Focus Array 1386935_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700014548.rn.1 TC0700001870.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014547.rn.1 TC0700001870.rn.1 RTA TC
          Rat Expression Set 230 1386935_at - -
          Rat Expression Set 230 1386935_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899387 - -
          Rat Genome U34 Set U17254_at - -
          Rat Genome U34 Set U17254_g_at - -
          Rat Genome U34 Set U17254_at - -
          Rat Genome U34 Set U17254_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20970853 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361354 - -
          RatRef-12 Expression BeadChip ILMN_63698 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline