Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn00668140_g1
See other CDH1 GE Assays ›
Gene Symbol:
Cdh1
Gene Name:
cadherin 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 38768467 - 38838395 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn00668140_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Cdh1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_031334.1
NP_112624.1
12 - 13
2080
84
GenBank mRNA
AB017696.1
-
12 - 13
2080
84
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1900000526.rn.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Detect Genomic DNA
Rattus norvegicus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Cdh1
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
trophectodermal cell differentiation
epithelial cell morphogenesis
homophilic cell adhesion via plasma membrane adhesion molecules
synapse assembly
sensory perception of sound
response to toxic substance
response to organic substance
single organismal cell-cell adhesion
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
protein metabolic process
pituitary gland development
negative regulation of cell-cell adhesion
neuron projection development
regulation of protein localization
regulation of water loss via skin
adherens junction organization
response to drug
positive regulation of transcription factor import into nucleus
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of transcription, DNA-templated
negative regulation of epithelial cell proliferation
protein homooligomerization
intestinal epithelial cell development
salivary gland cavitation
regulation of branching involved in salivary gland morphogenesis
bicellular tight junction assembly
cellular response to amino acid stimulus
cellular response to lithium ion
cellular response to indole-3-methanol
protein localization to plasma membrane
establishment of protein localization to plasma membrane
negative regulation of canonical Wnt signaling pathway
cochlea development
regulation of protein localization to cell surface
regulation of neuron migration
glycoprotein binding
calcium ion binding
protein binding
beta-catenin binding
protein phosphatase binding
protein domain specific binding
ankyrin binding
GTPase activating protein binding
gamma-catenin binding
cell adhesion molecule binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1900004594.rn.1
TC1900000526.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline