Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00668527_g1
          See other NR1H3 GE Assays ›
          Gene Symbol:
          Nr1h3
          Gene Name:
          nuclear receptor subfamily 1, group H, member 3
          Gene Aliases:
          LXRalpha
          Chromosome Location:
          Chr.3: 80004130 - 80014197 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00668527_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr1h3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031627.2 NP_113815.2 8 - 9 1212 73
          XM_006234533.2 XP_006234595.1 9 - 10 1390 73
          XM_006234534.1 XP_006234596.1 9 - 10 1331 73
          XM_006234535.2 XP_006234597.1 9 - 10 1532 73
          XM_006234537.1 XP_006234599.1 8 - 9 1002 73
          XM_008762013.1 XP_008760235.1 8 - 9 1122 73
          GenBank mRNA BC088750.1 - 8 - 9 1212 7314121 73
          U11685.1 - 8 - 9 1219 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003650.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr1h3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          lipid metabolic process
          insulin receptor signaling pathway
          positive regulation of triglyceride biosynthetic process
          positive regulation of receptor biosynthetic process
          positive regulation of cholesterol efflux
          negative regulation of cholesterol storage
          intracellular receptor signaling pathway
          positive regulation of cellular protein metabolic process
          negative regulation of lipid transport
          positive regulation of cholesterol transport
          response to progesterone
          positive regulation of toll-like receptor 4 signaling pathway
          cholesterol homeostasis
          negative regulation of macrophage activation
          apoptotic cell clearance
          steroid hormone mediated signaling pathway
          cellular lipid metabolic process
          positive regulation of fatty acid biosynthetic process
          negative regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of pinocytosis
          negative regulation of inflammatory response
          positive regulation of lipoprotein lipase activity
          lipid homeostasis
          sterol homeostasis
          triglyceride homeostasis
          cellular response to lipopolysaccharide
          negative regulation of pancreatic juice secretion
          negative regulation of secretion of lysosomal enzymes
          regulation of cholesterol homeostasis
          positive regulation of cholesterol homeostasis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          zinc ion binding
          sterol response element binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          retinoid X receptor binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140865 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12618 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12618 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12618 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700695623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22308 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69991 - -
          Agilent Rat cDNA Microarray (G4105A) 700695623 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10847355 - -
          Affymetrix Rat Gene 1.0 ST Array 10847359 - -
          Affymetrix Rat Gene 1.0 ST Array 10847356 - -
          Affymetrix Rat Gene 1.0 ST Array 10847360 - -
          Affymetrix Rat Gene 1.0 ST Array 10847353 - -
          Affymetrix Rat Gene 1.0 ST Array 10847361 - -
          Affymetrix Rat Gene 1.0 ST Array 10847358 - -
          Affymetrix Rat Gene 1.0 ST Array 10847354 - -
          Affymetrix Rat Gene 1.0 ST Array 10847352 - -
          Affymetrix Rat Gene 1.0 ST Array 10847362 - -
          Affymetrix Rat Gene 1.0 ST Array 10847357 - -
          Affymetrix Rat Gene 1.1 ST Array 10847351 - -
          Rat Genome 230 2.0 Array 1387365_PM_at - -
          Rat Genome 230 2.0 Array 1387365_at - -
          Affymetrix HT Rat Focus Array 1387365_at - -
          Affymetrix HT Rat Focus Array 1387365_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300027721.rn.1 TC0300003650.rn.1 RTA TC
          Rat Expression Set 230 1387365_at - -
          Rat Expression Set 230 1387365_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10847351 - -
          Rat Genome U34 Set U11685_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21852536 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352599 - -
          RatRef-12 Expression BeadChip ILMN_51603 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline