Search
Search

Slco1b2| Gene Symbol: Slco1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270586.1 | NP_001257515.1 | 11 - 12 | 1501 | 85 | |
| NM_001270587.1 | NP_001257516.1 | 10 - 11 | 1507 | 85 | ||
| NM_031650.3 | NP_113838.1 | 11 - 12 | 1606 | 85 | ||
| GenBank mRNA | AF147740.1 | - | 10 - 11 | 1387 | 85 | |
| AF208545.2 | - | 11 - 12 | 1606 | 85 | ||
| AF217450.2 | - | 10 - 11 | 1507 | 85 | ||
| AJ271682.1 | - | 11 - 12 | 1607 | 85 | ||
Set Membership: |
|