Search
Search
| Gene Symbol: Ppig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031793.1 | NP_113981.1 | 6 - 7 | 575 | 149 | |
| XM_006234324.2 | XP_006234386.1 | 6 - 7 | 951 | 149 | ||
| XM_006234325.2 | XP_006234387.1 | 6 - 7 | 772 | 149 | ||
| XM_006234326.2 | XP_006234388.1 | 6 - 7 | 895 | 149 | ||
| XM_006234327.2 | XP_006234389.1 | 5 - 6 | 847 | 149 | ||
| XM_006234328.2 | XP_006234390.1 | 6 - 7 | 842 | 149 | ||
| XM_006234330.2 | XP_006234392.1 | 5 - 6 | 668 | 149 | ||
| XM_006234331.2 | XP_006234393.1 | 5 - 6 | 794 | 149 | ||
| XM_006234333.2 | XP_006234395.1 | 5 - 6 | 808 | 149 | ||
| XM_006234334.2 | XP_006234396.1 | 5 - 6 | 629 | 149 | ||
| XM_006234335.2 | XP_006234397.1 | 5 - 6 | 755 | 149 | ||
| XM_008761930.1 | XP_008760152.1 | 5 - 6 | 871 | 149 | ||
| XM_008761931.1 | XP_008760153.1 | 5 - 6 | 832 | 149 | ||
| GenBank mRNA | AF043642.1 | - | 6 - 7 | 575 | 149 | |
| BC085723.1 | - | 5 - 6 | 621 | 7123868 | 149 | |
| FQ225838.1 | - | 1 - 2 | 90 | 149 | ||
| FQ230740.1 | - | 1 - 2 | 90 | 149 | ||
| FQ232477.1 | - | 1 - 2 | 90 | 149 | ||
Set Membership: |
|