Search
Search
| Gene Symbol: Kcnh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053937.1 | NP_446389.1 | 11 - 12 | 2425 | 57 | |
| XM_006247554.2 | XP_006247616.1 | 12 - 13 | 2516 | 57 | ||
| XM_006247555.2 | XP_006247617.1 | 12 - 13 | 2474 | 57 | ||
| XM_006247556.2 | XP_006247618.1 | 11 - 12 | 2453 | 57 | ||
| GenBank mRNA | AF016192.1 | - | 11 - 12 | 2425 | 57 | |
Set Membership: |
|