Search
Search
| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023020.2 | NP_075409.1 | 4 - 5 | 749 | 91 | |
| GenBank mRNA | AJ250730.1 | - | 4 - 5 | 546 | 91 | |
| BC129093.1 | - | 4 - 5 | 749 | 8363015 | 91 | |
Set Membership: |
|