Search
Search
| Gene Symbol: Gabre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023091.1 | NP_075579.1 | 5 - 6 | 2113 | 126 | |
| GenBank mRNA | AF189262.1 | - | 5 - 6 | 2113 | 126 | |
| AF255385.1 | - | 1675 | 126 | |||
| AF255386.1 | - | 3 - 4 | 368 | 126 | ||
| AF255387.1 | - | 3 - 4 | 368 | 126 | ||
| AF255612.1 | - | 678 | 126 | |||
Set Membership: |
|