Search
Search
| Gene Symbol: Gfra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023967.1 | NP_076457.1 | 3 - 4 | 592 | 75 | |
| XM_006235040.2 | XP_006235102.1 | 2 - 3 | 2065 | 75 | ||
| XM_006235041.2 | XP_006235103.1 | 2 - 3 | 2065 | 75 | ||
| XM_006235042.2 | XP_006235104.1 | 2 - 3 | 2065 | 75 | ||
| XM_006235043.2 | XP_006235105.1 | 2 - 3 | 2065 | 75 | ||
| XM_006235045.2 | XP_006235107.1 | 3 - 4 | 1505 | 75 | ||
| XM_008762196.1 | XP_008760418.1 | 3 - 4 | 1505 | 75 | ||
| XM_008762197.1 | XP_008760419.1 | 3 - 4 | 1505 | 75 | ||
| GenBank mRNA | AJ294475.1 | - | 3 - 4 | 592 | 75 | |
| AJ294476.1 | - | 3 - 4 | 592 | 75 | ||
Set Membership: |
|