Search
Search
| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031342.1 | NP_112632.1 | 8 - 9 | 556 | 103 | |
| GenBank mRNA | AB021645.1 | - | 8 - 9 | 556 | 103 | |
| BC070503.1 | - | 8 - 9 | 562 | 7096587 | 103 | |
| FQ219970.1 | - | 2 - 3 | 148 | 103 | ||
Set Membership: |
|