Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00676266_m1
          See other VDAC3 GE Assays ›
          Gene Symbol:
          Vdac3
          Gene Name:
          voltage-dependent anion channel 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 74292466 - 74308910 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00676266_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vdac3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031355.1 NP_112645.1 6 - 7 641 107
          XM_006253358.2 XP_006253420.1 7 - 8 644 107
          GenBank mRNA AB039664.1 - 6 - 7 641 107
          AF048829.1 - 5 - 6 525 107
          AF048830.1 - 5 - 6 528 107
          AF268469.1 - 5 - 6 552 107
          BC061780.1 - 6 - 7 577 5623339 107
          FQ215155.1 - 1 - 2 228 107
          FQ215672.1 - 1 - 2 228 107
          FQ215800.1 - 1 - 2 227 107
          FQ216638.1 - 7 - 8 629 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000874.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vdac3

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          anion transport
          chemical synaptic transmission
          neuron-neuron synaptic transmission
          learning
          regulation of anion transport
          transmembrane transport
          regulation of cilium assembly
          nucleotide binding
          voltage-gated anion channel activity
          porin activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88846 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110242 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064187 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064187 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064187 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1042387 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700437159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88524 - -
          Agilent Rat cDNA Microarray (G4105A) 700437159 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789136 - -
          Affymetrix Rat Gene 1.0 ST Array 10789128 - -
          Affymetrix Rat Gene 1.0 ST Array 10789129 - -
          Affymetrix Rat Gene 1.0 ST Array 10789135 - -
          Affymetrix Rat Gene 1.0 ST Array 10789137 - -
          Affymetrix Rat Gene 1.0 ST Array 10789132 - -
          Affymetrix Rat Gene 1.0 ST Array 10789134 - -
          Affymetrix Rat Gene 1.0 ST Array 10789130 - -
          Affymetrix Rat Gene 1.0 ST Array 10789133 - -
          Affymetrix Rat Gene 1.0 ST Array 10789131 - -
          Affymetrix Rat Gene 1.1 ST Array 10789127 - -
          Rat Genome 230 2.0 Array 1386902_at - -
          Rat Genome 230 2.0 Array 1386902_PM_at - -
          Affymetrix HT Rat Focus Array 1386902_PM_at - -
          Affymetrix HT Rat Focus Array 1386902_at - -
          Affymetrix Rat Clariom S Assay PSR1600006741.rn.1 TC1600000874.rn.1 RTA TC
          Rat Expression Set 230 1386902_at - -
          Rat Expression Set 230 1386902_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789127 - -
          Rat Genome U34 Set rc_AI236370_f_at - -
          Rat Genome U34 Set rc_AA817975_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21078162 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367494 - -
          RatRef-12 Expression BeadChip ILMN_65818 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline