Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00677722_g1
          See other CDK4 GE Assays ›
          Gene Symbol:
          Cdk4
          Gene Name:
          cyclin-dependent kinase 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 70345971 - 70352689 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00677722_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdk4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053593.2 NP_446045.1 7 - 8 1007 65
          GenBank mRNA BC070956.1 - 7 - 8 1007 7104124 65
          FQ214013.1 - 7 - 8 1023 65
          FQ234663.1 - 6 - 7 636 65
          L11007.1 - 7 - 8 939 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000886.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdk4

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          lens development in camera-type eye
          protein phosphorylation
          signal transduction
          circadian rhythm
          positive regulation of cell proliferation
          response to toxic substance
          response to organic substance
          response to lead ion
          regulation of gene expression
          positive regulation of G2/M transition of mitotic cell cycle
          organ regeneration
          response to testosterone
          regulation of cell proliferation
          response to drug
          positive regulation of apoptotic process
          positive regulation of translation
          positive regulation of cell size
          positive regulation of fibroblast proliferation
          cell division
          regulation of cell cycle
          response to hyperoxia
          negative regulation of cell cycle arrest
          protein kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          ATP binding
          kinase activity
          cyclin binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P388782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154478 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P388782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P388782 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P388782 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P388777 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P388782 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700148320 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700547120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97001 - -
          Agilent Rat cDNA Microarray (G4105A) 700547120 - -
          Agilent Rat cDNA Microarray (G4105A) 700148320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895802 - -
          Affymetrix Rat Gene 1.0 ST Array 10895801 - -
          Affymetrix Rat Gene 1.0 ST Array 10895800 - -
          Affymetrix Rat Gene 1.0 ST Array 10895798 - -
          Affymetrix Rat Gene 1.0 ST Array 10895799 - -
          Affymetrix Rat Gene 1.1 ST Array 10895797 - -
          Rat Genome 230 2.0 Array 1372739_at - -
          Rat Genome 230 2.0 Array 1369950_PM_at - -
          Rat Genome 230 2.0 Array 1372739_PM_at - -
          Rat Genome 230 2.0 Array 1369950_at - -
          Affymetrix HT Rat Focus Array 1369950_at - -
          Affymetrix HT Rat Focus Array 1369950_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700006602.rn.1 TC0700000886.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700006603.rn.1 TC0700000886.rn.1 RTA TC
          Rat Expression Set 230 1369950_PM_at - -
          Rat Expression Set 230 1369950_at - -
          Affymetrix Rat Gene 1.0 ST Array 10895797 - -
          Rat Genome U34 Set L11007_at - -
          Rat Genome U34 Set L11007_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21324626 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373114 - -
          RatRef-12 Expression BeadChip ILMN_58091 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline