Search
Search
| Gene Symbol: Arfrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053980.1 | NP_446432.1 | 5 - 6 | 486 | 66 | |
| XM_006235718.2 | XP_006235780.1 | 5 - 6 | 482 | 66 | ||
| GenBank mRNA | X78603.1 | - | 5 - 6 | 486 | 66 | |
Set Membership: |
|