Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00678963_g1
          See other NSMF GE Assays ›
          Gene Symbol:
          Nsmf
          Gene Name:
          NMDA receptor synaptonuclear signaling and neuronal migration factor
          Gene Aliases:
          Jac, Jacob, Nelf
          Chromosome Location:
          Chr.3: 2261950 - 2270996 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00678963_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nsmf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270626.1 NP_001257555.1 14 - 15 1636 90
          NM_001270627.1 NP_001257556.1 13 - 14 1546 90
          NM_001270628.1 NP_001257557.1 13 - 14 1630 90
          NM_057190.2 NP_476538.2 15 - 16 1705 90
          NR_073057.1 14 - 15 1580 90
          XM_006233554.2 XP_006233616.1 14 - 15 1918 90
          XM_008761575.1 XP_008759797.1 14 - 15 2002 90
          XM_008761576.1 XP_008759798.1 13 - 14 1912 90
          GenBank mRNA AJ293697.1 - 15 - 16 1638 90
          AJ293698.1 - 14 - 15 1569 90
          AJ293699.1 - 13 - 14 1479 90
          AJ534640.1 - 13 - 14 1562 90
          AJ534642.1 - 14 - 15 1512 90
          BC087719.1 - 15 - 16 1666 7318793 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000034.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nsmf

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          positive regulation of protein dephosphorylation
          regulation of neuron apoptotic process
          regulation of neuronal synaptic plasticity
          regulation of dendrite morphogenesis
          cellular response to amino acid stimulus
          cellular response to electrical stimulus
          cellular response to gonadotropin stimulus
          positive regulation of neuron migration
          molecular_function
          protein binding
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042380 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P494147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90202 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042380 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P494147 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P494147 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042380 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P494147 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042380 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P494147 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21011 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834562 - -
          Affymetrix Rat Gene 1.0 ST Array 10834564 - -
          Affymetrix Rat Gene 1.0 ST Array 10834551 - -
          Affymetrix Rat Gene 1.0 ST Array 10834566 - -
          Affymetrix Rat Gene 1.0 ST Array 10834565 - -
          Affymetrix Rat Gene 1.0 ST Array 10834563 - -
          Affymetrix Rat Gene 1.0 ST Array 10834557 - -
          Affymetrix Rat Gene 1.0 ST Array 10834561 - -
          Affymetrix Rat Gene 1.0 ST Array 10834554 - -
          Affymetrix Rat Gene 1.0 ST Array 10834550 - -
          Affymetrix Rat Gene 1.0 ST Array 10834555 - -
          Affymetrix Rat Gene 1.0 ST Array 10834558 - -
          Affymetrix Rat Gene 1.0 ST Array 10834559 - -
          Affymetrix Rat Gene 1.0 ST Array 10834552 - -
          Affymetrix Rat Gene 1.0 ST Array 10834553 - -
          Affymetrix Rat Gene 1.0 ST Array 10834560 - -
          Affymetrix Rat Gene 1.0 ST Array 10834556 - -
          Affymetrix Rat Gene 1.1 ST Array 10834549 - -
          Rat Genome 230 2.0 Array 1368153_a_at - -
          Rat Genome 230 2.0 Array 1368153_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368153_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368153_a_at - -
          Affymetrix Rat Clariom S Assay PSR0300000269.rn.1 TC0300000034.rn.1 RTA TC
          Rat Expression Set 230 1368153_PM_a_at - -
          Rat Expression Set 230 1368153_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834549 - -
          Rat Genome U34 Set rc_AA943103_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21817538 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351055 - -
          RatRef-12 Expression BeadChip ILMN_56446 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline