Search
Search

Bpifa2f| Gene Symbol: Bpifa2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080775.1 | NP_542953.1 | 4 - 5 | 478 | 82 | |
| XM_008762380.1 | XP_008760602.1 | 4 - 5 | 590 | 82 | ||
| GenBank mRNA | M83210.1 | - | 4 - 5 | 478 | 82 | |
Set Membership: |
|